Search Thermo Fisher Scientific
MagMAX kits and reagents use magnetic bead-based technology to purify high-quality DNA and RNA from cell-free plasma, serum, or urine samples. To help save time and increase reproducibility, combine with KingFisher instruments for automated purification.
MagMAX kits purify high-quality nucleic acids, cell-free samples, and FFPE blocks, and are suitable for a range of research applications, including next-generation sequencing (NGS), qPCR, and digital PCR.
MagMAX Cell-Free Total Nucleic Acid Isolation Kit Purify free-circulating DNA, RNA and miRNA | MagMAX Cell-Free DNA Isolation Kit Isolate cell-free DNA in | MagMAX FFPE DNA/RNA Ultra Kit Sequential isolation of | |
---|---|---|---|
![]() | ![]() | ![]() | |
Sample volumes | 1.0–6 mL plasma | 100 µL–10 mL | 5–40 µm standard input of slides or curls |
Elution volumes | 15–60 μL | 15–50 µL | 20–50 µL |
Time per run | 90 minutes total, 10–15 min hands-on time with KingFisher instruments | 45–60 min | 20 min for DNA, 48 min for RNA |
Automation/HT |
|
|
|
— | Learn more | Learn more | |
Order now | Order now | Order now |
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.