Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Toxicity is crucial in biological research and plays an important role in drug development. Toxicity studies aim to remove toxic compounds early in the drug development process, as drug-induced toxicities are the leading cause of drug failures.
Organ-specific toxicity tests—specifically the kidney—are of high interest as a central detoxification organ for their high exposure to drugs, reactive metabolites, and environmental chemicals. Kidney toxicity is routinely assessed during preclinical safety evaluations.
Invitrogen ProcartaPlex immunoassays for kidney toxicity enable the simultaneous quantification of up to 11 targets in a single well. These assays are available as single targets, preconfigured panels of targets, and custom-blended Mix & Match panels.
Simultaneous detection of these targets can help to gain more information about kidney functions and nephrotoxicity during drug development studies.
ProcartaPlex Simplex kits are designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits, only one buffer kit #EPX010-10420-901 (sold separately) is needed for each assay plate regardless of plex size.
View the complete list of ProcartaPlex panels
Custom-blended panels deliver results tailored to your choice of targets. Simply provide your desired species, sample type, and instrument for use. Then select from our list of analytes and we will formulate a user-defined panel that is supplied with all the necessary reagents to perform the assay.
Multiplex up to 80 RNA targets from a single biological sample.
Explore the different Luminex systems: Luminex 200, FLEXMAP3D, and INTELLIFLEX models, which are all compatible with Invitrogen Luminex Assays.
Detect very low levels of proteins using as little as 1–5 μL of sample.
Quantitate single analytes with confidence with highly verified Invitrogen ELISA kits.
*FDA guidance for nephrotoxicity biomarkers: https://www.fda.gov/media/135310/download and https://www.fda.gov/media/87781/download
For Research Use Only. Not for use in diagnostic procedures.