Search Thermo Fisher Scientific
Invitrogen ProcartaPlex Preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide optimal performance. Precision and lot-to-lot consistency is guaranteed by highly validated analyte combinations, ensuring reproducibility during longitudinal studies.
ProcartaPlex Preconfigured panels are predefined mixtures of target-specific magnetic beads, detection antibodies, and protein standards for quantitative analysis of up to 80 targets in a single sample. Panels are available for samples from six species including human, mouse, rat, non-human primate, porcine, and canine.
Use the tabs below to browse the available panels by species.
Human multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Human panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Immune Response 80-Plex Human Panel | EPX800-10080-901 | 80 | APRIL, BAFF, BLC, bNGF, CCL1 (I-309), CCL17 (TARC), CCL21 (6Ckine/SLC), CCL23 (MPIF), CCL25 (TECK), CD30, CD40L, CXCL6 (GCP-2), ENA-78, Eotaxin, Eotaxin-2, Eotaxin-3, FGF-2, Fractalkine, Gal-3, G-CSF, GM-CSF, Granzyme A, Granzyme B, Gro a, HGF, IFN-a, IFN-g, IL-10, IL-12p70, IL-13, IL-15, IL-16, IL-17A, IL-18, IL-1a, IL-1b, IL-2, IL-20, IL-21, IL-22, IL-23, IL-27, IL-2R, IL-3, IL-31, IL-34, IL-37 , IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IP-10, I-TAC, LIF, MCP-1, MCP-2, MCP-3, MCP-4 (CCL13), M-CSF, MDC, MIF, MIG, MIP1a, MIP-1b, MIP-2 alpha (CXCL2), MIP-3 beta (CCL19), MIP-3a, MMP-1, PTX3, SCF, TNF b, TNF-a, TNF-R2, TRAIL, TREM-1, TSLP, TWEAK, VEGF-A |
Immune Monitoring 65-Plex Human Panel | EPX650-10065-901 | 65 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-16, IL-17A (CTLA-8), IL-18, IL-20, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, M-CSF, MIF, TNF alpha, TNF beta, TSLP, BLC (CXCL13), ENA-78 (CXCL5), Eotaxin (CCL11), Eotaxin-2 (CCL24), Eotaxin-3 (CCL26), Fractalkine (CX3CL1), Gro-alpha (CXCL1), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (CCL7), MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-3 alpha (CCL20), SDF-1 alpha (CXCL12), FGF-2, HGF, MMP-1, NGF beta, SCF, VEGF-A, APRIL, BAFF, CD30, CD40L (CD154), IL-2R (CD25), TNF-RII, TRAIL (CD253), TWEAK |
Cytokine/Chemokine/Growth Factor Convenience 45-Plex Human Panel 1 | EPXR450-12171-901 | 45 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, SCF, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, BDNF, EGF, FGF-2, HGF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Immune Checkpoint 37-Plex Human Panel | EPX370-15846-901 | 37 | Arginase-1, B7-H6, BTLA, CD134 (OX40), CD137 (4-1BB), CD152 (CTLA4), CD27, CD276 (B7-H3), CD28, CD47 (IAP), CD48 (BLAST-1), CD73 (NT5E), CD80, CD96 (Tactile), E-cadherin, GITR, HVEM, ICOS ligand (B7-H2), IDO, LAG-3, MICA, MICB, nectin-2, PD-1, PD-L1, PD-L2, perforin, PVR, S100A8/A9, siglec-7, siglec-9, TIM-3, TIMD-4, ULBP-1, ULBP-3, ULBP-4, VISTA (B7-H5) |
Cytokine & Chemokine Convenience 34-Plex Human Panel 1A | EPXR340-12167-901 | 34 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Abundant Serum Markers 26-Plex Human Panel | EPX260-15808-901 | 26 | Adiponectin, CD14, CD44, CD44var (v6), CD62L (L-Selectin), Cystatin C, Elafin, Endoglin, ICAM-1, Lactoferrin, Leptin, MBL, MMP-3, MMP-9, NGAL, NRP-1, Osteopontin (OPN), PAI-1 (Serpin), RANTES (CCL5), RBP4, REG-3a, Resistin (ADSF), SCGF beta, TIMP-1, VCAM-1, ZAG |
Cytokine Storm 21-Plex Human Panel | EPX210-15850-901 | 21 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), TNF alpha, TNF beta |
Inflammation 20-Plex Human Panel | EPX200-12185-901 | 20 | CD62P (P-Selectin), CD62E (E-selectin), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IP-10 (CXCL10), ICAM-1, MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), TNF alpha |
Angiogenesis 18-Plex Human Panel | EPX180-15806-901 | 18 | Angiopoietin-1, BMP-9, CD31 (PECAM-1), EGF, EMMPRIN, Follistatin, FGF-2, G-CSF (CSF-3), HB-EGF, HGF, IL-8 (CXCL8), Leptin, LYVE-1, PDGF-BB, Syndecan, TIE-2, VEGF-A, VEGF-D |
Neuroscience 18-plex Human Panel | EPX180-15837-901 | 18 | Amyloid beta 1-42, BDNF, YKL-40 (CHI3L1), CNTF, FGF-21, GDNF, GFAP, Kallikrein-6 (KLK6), MIF, NCAM-1, Neurogranin (NRGN), NF-H, NGF beta, S100B, Tau (total), Tau (pT181), TDP-43, UCHL1 |
TNF Superfamily 15-Plex Human Panel | EPX150-15801-901 | 15 | CD137L (4-1BBL), APRIL, BAFF, CD30L, CD40L, EDA-1, Fas-L, GITRL, LIGHT (CD258), NGF beta, RANKL, TNF alpha, TNF beta, TRAIL, TWEAK |
Immuno-Oncology Checkpoint 14-Plex Human Panel 1 | EPX14A-15803-901 | 14 | BTLA, CD80, CD152 (CTLA4), CD27, CD28, CD137 (4-1BB), GITR, HVEM, IDO, LAG-3, PD-1, PD-L1, PD-L2, TIM-3 |
Immuno-Oncology Checkpoint 14-Plex Human Panel 2 | EPX140-15815-901 | 14 | Arginase-1, CD73 (NT5E), CD96 (Tactile), E-Cadherin, Nectin-2, MICA, MICB, PVR, Siglec-7, Perforin, Siglec-9, ULBP-1, ULBP-3, ULBP-4 |
Cell Proliferation and Metastasis 13-Plex Human Panel 1 | EPX130-15841-901 | 13 | AFP (alpha-fetoprotein), ADAM12, beta-catenin, CD171 (NCAM-L1), CD66f (PSBG-1), galectin-1, hCG, HO-1 (heme oxygenase 1), IGFBP-1, N-cadherin, nectin-4, PAPP-A, thrombomodulin (TM) 1 |
Apoptotic Cell Clearance 12-Plex Human Panel | EPX120-15816-901 | 12 | AXL, CD36, LOX-1, Mer (MERTK), CD31 (PECAM-1), RAGE, TYRO3, Gas6, MBL ,uPAR, CALR (CRT), PAI-1 (Serpin) |
Cell Proliferation and Metastasis 12-Plex Human Panel 2 | EPX120-15842-901 | 12 | beta-2-microglobulin (B2M), cathepsin D, CEA (CEACAM-5), EGFR (ErbB1), haptoglobin, HGFR (c-Met), IGFBP-2, IGFBP-3, MIA, MIP-4 (CCL18), periostin (OSF-2), VE-cadherin |
Endothelial Injury Marker 12-Plex Human Panel | EPX120-15849-901 | 12 | E-selectin, P-selectin, VEGF-A, VEGF-D, PECAM-1, Angiopoietin-1, Syndecan-1, VEGF-R2, VEGF-R3, MMP-1, Thrombomodulin (TM), VLA-4 |
TNF Receptor Superfamily 12-Plex Human Panel | EPX120-15802-901 | 12 | CD27, CD30, CD40, CD137 (4-1BB), DR6, Fas (APO), GITR, HVEM, TNF-RI, TNF-RII, TRAIL-R1, TRAIL-R2 |
Coronavirus Ig Total Human 11-Plex Panel | EPX110-16000-901 | 11 | SARS-CoV-2 Spike (trimer), SARS-CoV-2 S1 protein, SARS-CoV-2 RBD (IAP), SARS-CoV-2 Nucleocapsid, SARS S1, CoV-NL63, CoV-HKU1, CoV-229E, MERS S1, CoV-OC43, Negative control |
Kidney Toxicity Panel 1 | EPX110-15855-901 | 11 | KIM-1 (TIM-1/HAVCR1), Renin, Calbindin, Osteoactivin (GPNMB), GSTA1 (Glutathione S-transferase A1), IL-18, IP-10 (CXCL10), MCP-1 (CCL2), VEGF-A, Clusterin (APO-J), RBP4 (Retinol-binding protein 4) |
Kidney Toxicity Panel 2 | EPX090-15856-901 | 9 | Uromodulin, Alpha-1-Microglobulin, TFF3 (Trefoil Factor 3), Osteopontin (OPN), Cystatin C, NGAL, Beta-2-microglobulin (B2M), TIMP-1, EGF |
Atherosclerosis 9-Plex Human Panel 1 | EPX090-15822-901 | 9 | FGF-2, HSP60, IL-6, IL-8 (CXCL8), LOX-1, Myeloperoxidase (MPO), PAI-1 (Serpin), Tenascin-C, TRAIL |
Chemokine 9-Plex Human Panel 2 | EPX090-15840-901 | 9 | CCL1 (I-309), CCL17 (TARC), CCL21 (6Ckine/SLC), CCL23 (MPIF), CCL25 (TECK), CXCL6 (GCP-2), MCP-4 (CCL13), MIP-2 alpha (CXCL2), MIP-3 beta (CCL19) |
Immuno-Oncology Checkpoint 9-Plex Human Panel 3 | EPX090-15820-901 | 9 | B7-H6, CD276 (B7-H3), CD47 (IAP), CD48 (BLAST-1), CD134 (OX40), ICOS Ligand (B7-H2), TIMD-4, S100A8/A9, VISTA (B7-H5) |
Neurodegeneration 9-plex Human Panel 1 | EPX090-15836-901 | 9 | Amyloid beta 1-40, Amyloid beta 1-42, FGF-21, Kallikrein-6 (KLK-6), NCAM-1, Neurogranin (NRGN), Tau (total), Tau (pT181), TDP-43 |
Cardiac Remodeling 8-Plex Human Panel | EPX080-15819-901 | 8 | Galectin-3, IL-33, IL-33R (ST2), IP-10 (CXCL10), LAP, Osteopontin (OPN), PTX3, TREM-1 |
Cell Proliferation 8-Plex Human Panel | EPX080-15844-901 | 8 | EpCAM, glypican-1 (GPC1), HSP27, HSP60, IGF-2, midkine (MK), SPARC (osteonectin), TrkB |
Graft Versus Host Disease 8-Plex Human Panel | EPX080-15814-901 | 8 | CD30, Granzyme A, HGF, IL-2R, IL-33R (ST2), KRT18, Syndecan, TNF-RI |
Myokine 8-Plex Human Panel | EPX080-12186-901 | 8 | BDNF, IL-1RA, IL-10, IL-15, IL-8 (CXCL8), IL-6, LIF, TNF alpha |
Antibody Isotyping 7-Plex Human Panel | EPX070-10818-901 | 7 | IgG1, IgG2, IgG3, IgG4, IgA, IgE, IgM |
Cardiac Disease 7-Plex Human Panel | EPX070-15809-901 | 7 | Cardiotrophin-1 (CTF-1), CKMB, FABP-3, GDF-15, myoglobin, NTproBNP, troponin I (TNI) |
Apoptosis 6-Plex Human Panel | EPX06A-15823-901 | 6 | Bcl-2, Caspase-3 (active), cleaved PARP, Cytochrome C, GAPDH, p53 |
Coagulation 6-Plex Human Panel 1 | EPX060-10824-901 | 6 | Antithrombin, CRP, Factor XI, Factor XII, Factor XIII, Prothrombin |
Essential Th1/Th2 Cytokine 6-Plex Human Panel | EPX060-10009-901 | 6 | IFN gamma, IL-12p70, IL-4, IL-5, IL-6, TNF alpha |
Exosome Characterization 6-Plex Human Panel | EPX060-15845-901 | 6 | CD9, CD63, CD81, cytochrome c, syntenin-1, VLA-4 |
SARS-CoV-2 Variants Neutralizing Antibody 6-Plex Panel | EPX060-16018-901 | 6 | SARS-CoV-2 S1 B.1.1.529 (o), SARS-CoV-2 S1 B.1.617.2 (δ), SARS-CoV-2 Spike B.1.617.2 P.1 (γ), SARS-CoV-2 S1 B.1.351 (β), SARS-CoV-2 S1 B.1.1.7 (α), SARS-CoV-2 S1 (wt) |
Apolipoprotein 5-Plex Human Panel | EPX050-15818-901 | 5 | APO A1, APO A2, APO B, APO C2, APO E |
Arteriosclerosis 5-Plex Human Panel | EPX050-15821-901 | 5 | BMP-2, COMP, FGF-23, Osteoprotegerin (OPG), RANKL |
MMP 5-Plex Human Panel 1 | EPX050-10015-901 | 5 | MMP-1, MMP-7, MMP-8, MMP-12, MMP-13 |
Atherosclerosis 4-Plex Human Panel 2 | EPX040-15824-901 | 4 | Lp-PLA2, YKL-40 (CHI3L1), ICAM-1, Adiponectin |
Brain Injury 4-plex Human Panel | EPX040-15827-901 | 4 | NF-H, GFAP, UCHL1, S100B |
Cell Death 4-Plex Human Panel | EPX040-15843-901 | 4 | Dkk-1, HMGB1 (HMG-1), HSP70, HSP90 |
Coagulation 4-Plex Human Panel 3 | EPX040-10825-901 | 4 | Factor IX, Protein C, Protein S, vWF |
Essential 4-Plex Human Panel 2 | EPX040-10008-901 | 4 | IL-1 beta, IL-10, IL-17A (CTLA-8), IL-8 (CXCL8) |
Neurodegeneration 4-plex Human Panel 2 | EPX040-15832-901 | 4 | Apolipoprotein E4, Clusterin (Apo J), Complement Factor H, Fetuin-A |
Neurotrophic Factors 4-plex Human Panel | EPX040-15828-901 | 4 | GDNF, CNTF, BDNF, NGF beta |
Angiogenesis 3-Plex Human Panel 2 | EPX030-15807-901 | 3 | Angiogenin, Angiostatin, Endostatin |
MMP 3-Plex Human Panel 2 | EPX03A-10829-901 | 2 | MMP-2, MMP-3, MMP-9 |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Human Panel Name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 45-Plex Human Panel 1 | EPX450-12171-901 | 45 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, SCF, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, BDNF, EGF, FGF-2, HGF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Cytokine Chemokine 34-Plex Human Panel 1A | EPX340-12167-901 | 34 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Cytokine 25-Plex Human Panel 1B | EPX250-12166-901 | 25 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta |
Th1/Th2 & Chemokine 20-Plex Human Panel 1 | EPX200-12173-901 | 20 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8, IL-12p70, IL-13, IL-18, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, TNF alpha |
Th1/Th2 & Cytokine 18-Plex Human Panel 1C | EPX180-12172-901 | 18 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-12p70, IL-13, IL-15, IL-18, IL-31, TNF alpha, TNF beta |
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 18-Plex Human Panel | EPX180-12165-901 | 18 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, TNF alpha |
Th9/Th17/Th22/Treg & Chemokine 16-Plex Human Panel 1 | EPX160-12175-901 | 16 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-8 (CXCL8), IL-9, IL-10, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Cytokine 1C & Chemokine 16-Plex Human Panel 1 | EPX160-12176-901 | 16 | Eotaxin (CCL11), GRO alpha (CXCL1), IFN alpha, IL-1 alpha, IL-1RA, IL-7, IL-8 (CXCL8), IL-15, IL-31, GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, TNF beta |
Th9/Th17/Th22/Treg & Cytokine 14-Plex Human Panel 1C | EPX140-12174-901 | 14 | IFN alpha, IL-1 alpha, IL-1RA, IL-7, IL-9, IL-10, IL-15, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IL-31, TNF beta |
Growth Factor 11-Plex Human Panel | EPX110-12170-901 | 11 | BDNF, EGF, FGF-2, HGF, LIF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Th1/Th2 Cytokine 11-Plex Human Panel | EPX110-10810-90 | 11 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha |
Chemokine 9-Plex Human Panel 1 | EPX090-12187-901 | 9 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-8 (CXCL8), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha, RANTES (CCL5) |
Th9/Th17/Th22/Treg Cytokine 7-Plex Human Panel | EPX070-10817-901 | 7 | IL-10, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IL-9 |
Cytokine 7-Plex Human Panel 1C | EPX070-10010-90 | 7 | IFN alpha, IL-31, IL-15, IL-1 alpha, IL-1RA, IL-7, TNF beta |
The ProcartaPlex high-sensitivity immunoassays measure protein targets significantly below the detection limit of conventional assays for Luminex systems. Detect and quantify key cytokines that are expressed in the low pg/mL range.
HS panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
9-Plex Human Panel, High Sensitivity | EPXS090-12199-901 | 9 | IFN gamma, IL-1 beta, IL-2, IL-4, IL-6, IL-10, IL-12p70, IL-17A (CTLA-8), TNF alpha |
Mouse multiplex preconfigured panels are provided in a ready to use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Mouse panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Immune Response 64-Plex Mouse Panel | EPX640-20064-901 | 64 | BAFF, BLC (CXCL13), BTC, CD27, CTACK (CCL27), ENA-78 (CXCL5), Eotaxin (CCL11), Eotaxin-2 (CCL24), G-CSF (CSF-3), GM-CSF, Granzyme B, GRO alpha (CXCL1), IFN alpha, IFN gamma, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-16, IL-17A (CTLA-8), IL-18, IL-19, IL-1 alpha, IL-1 beta, IL-2, IL-22, IL-23, IL-25 (IL-17E), IL-27, IL-28, IL-2Ra, IL-3, IL-31, IL-33, IL-33R (ST2), IL-4, IL-5, IL-6, IL6R/sIL-6R, IL-7, IL-7R alpha, IL-9, IP-10 (CXCL10), I-TAC (CXCL11), Leptin, LIF, MCP-1 (CCL2), MCP-3 (CCL7), MCP-5 (CCL12), M-CSF, MDC (CCL22), MIP-1 alpha, MIP-1 beta, MIP-2, MIP-3b (CCL19), RANKL, RANTES (CCL5), SYB16 (CXCL16), TARC (CCL17), TECK (CCL25), TNF alpha, TSLP, VEGF-A, VEGF-R2 (KDR) |
Immune Monitoring 48-Plex Mouse Panel | EPX480-20834-901 | 48 | BAFF, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-19, IL-22, IL-23, IL-25 (IL-17E), IL-27, IL-28, IL-31, IL-33, LIF, M-CSF, RANKL, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), Betacellulin (BTC), Leptin, VEGF-A, IL-2R, IL-7R alpha, IL-33R (ST2) |
Cytokine & Chemokine Convenience 36-Plex Mouse Panel 1A | EPXR360-26092-901 | 36 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, LIF, M-CSF, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Cytokine & Chemokine Convenience 26-Plex Mouse Panel 1 | EPXR260-26088-901 | 26 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IP-10 (CXCL10), IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), TNF alpha |
Antibody Isotyping 7-Plex Mouse Panel 2 | EPX070-20816-901 | 7 | IgG1, IgG2b, IgG2c, IgG3, IgA, IgE, IgM |
Antibody Isotyping 7-Plex Mouse Panel | EPX070-20815-901 | 7 | IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, IgM |
Immuno-Oncology Checkpoint 7-Plex Mouse Panel 2 | EPX070-20835-901 | 7 | CD137L (4-1BBL), CD152 (CTLA4), CD276 (B7-H3), CD80, PD-1, PD-L1 |
Myokine 5-Plex Mouse Panel | EPX050-20833-901 | 5 | IL-10, IL-15/IL-15R, IL-6, LIF, TNF alpha |
Immuno-Oncology Checkpoint 4-Plex Mouse Panel 1 | EPX040-20830-901 | 4 | TIM-3, LAG-3, BTLA, CD27 |
IFN alpha/IFN beta 2-Plex Mouse Panel | EPX02A-22187-901 | 2 | IFN alpha, IFN beta |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Mouse panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 36-Plex Mouse Panel 1A | EPX360-26092-901 | 36 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, LIF, M-CSF, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Cytokine & Chemokine 26-Plex Mouse Panel 1 | EPX260-26088-901 | 26 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IP-10 (CXCL10), IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), TNF alpha |
Th1/Th2 & Chemokine 20-Plex Mouse Panel 1 | EPX200-26090-901 | 20 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha, IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 17-Plex Mouse Panel | EPX170-26087-901 | 17 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, TNF alpha |
Th9/Th17/Th22/Treg & Chemokine 15-Plex Mouse Panel | EPX150-26089-901 | 15 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-9, IL-10, IL-17A (CTLA-8), IL-22, IL-23, IL-27, IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Th1/Th2 Cytokine 11-Plex Mouse Panel | EPX110-20820-901 | 11 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha |
Cytokine 10-Plex Mouse Panel 1B | EPX100-26091-901 | 10 | ENA-78 (CXCL5), G-CSF (CSF-3), IFN alpha, IL-1 alpha, IL-3, IL-15/IL-15R, IL-28, IL-31, LIF, M-CSF |
Chemokine 9-Plex Mouse Panel 1 | EPX090-20821-901 | 9 | Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Essential 8-Plex Mouse Panel 2 | EPX080-20832-901 | 8 | GRO alpha (CXCL1), IL-1 beta, IL-10, IL-17A (CTLA-8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2 |
Th9/Th17/Th22/Treg Cytokine 6-Plex Mouse Panel | EPX060-20822-901 | 6 | IL-10, IL-17A (CTLA-8), IL-22, IL-23, IL-27, IL-9 |
Essential Th1/Th2 Cytokine 6-Plex Mouse Panel | EPX060-20831-901 | 6 | IFN gamma, IL-12p70, IL-4, IL-5, IL-6, TNF alpha |
The ProcartaPlex high-sensitivity immunoassays measure protein targets significantly below the detection limit of conventional assays for Luminex systems. Detect and quantify key cytokines that are expressed in the low pg/mL range.
HS panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
5-Plex Mouse Panel, High Sensitivity | EPXS050-22199-901 | 5 | IFN gamma, IL-2, IL-4, IL-6, TNF alpha |
Rat multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex Assays.
Rat panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Antibody Isotyping 6-Plex Rat Panel | EPX060-30123-901 | 6 | IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM |
EPX050-30124-901 | 5 | Calbindin, KIM-1, Osteopontin (OPN), TFF3, VEGF-A | |
EPX050-30125-901 | 5 | Albumin, Clusterin (Apo-J), Cystatin, NGAL, TIMP-1 |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Rat panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 22-Plex Rat Panel | EPX220-30122-901 | 22 | Eotaxin (CCL11), GRO alpha (CXCL1), G-CSF (CSF-3), GM-CSF, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A(CTLA-8), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-2, RANTES (CCL5), TNF alpha |
Th Complete 14-Plex Rat Panel | EPX140-30120-901 | 14 | G-CSF (CSF-3), GM-CSF, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), TNF alpha |
Chemokine 8-Plex Rat Panel | EPX080-30121-901 | 8 | Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-2, RANTES (CCL5) |
Canine or Porcine multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex Assays.
Canine panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 11-Plex Canine Panel 1 | EPX11A-50511-901 | 11 | IFN gamma, IL-2, IL-6, IL-8 (CXCL8), IL-10, IL-12/IL-23p40, MCP-1 (CCL2), NGF beta, SCF, TNF alpha, VEGF-A |
Porcine panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 9-Plex Porcine Panel 1 | EPX090-60829-901 | 9 | IFN alpha, IFN gamma, IL-1 beta, IL-10, IL-12/IL-23p40, IL-4, IL-6, IL-8 (CXCL8), TNF alpha |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50x) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Non-human primate panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 37-Plex NHP Panel | EPX370-40045-901 | 37 | CD40L, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-23, SCF, TNF alpha, BLC, Eotaxin (CCL11), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha, BDNF, FGF-2, NGF beta, PDGF-BB, SCF, VEGF-A, VEGF-D |
Cytokine & Chemokine 30-Plex NHP Panel | EPX300-40044-901 | 30 | CD40L, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-23, TNF alpha, BLC (CXCL13), Eotaxin (CCL11), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha |
Th 14-Plex NHP Panel | EPX140-40040-901 | 14 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-23, TNF alpha |
Chemokine 10-Plex NHP Panel | EPX100-40041-901 | 10 | BLC (CXCL13), Eotaxin (CCL11), IL-8 (CXCL8), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha |
Growth Factor 7-Plex NHP Panel | EPX070-40043-901 | 7 | BDNF, FGF-2, NGF beta, PDGF-BB, SCF, VEGF-A, VEGF-D |
Cytokine 6-Plex NHP Panel 1B | EPX060-40042-901 | 6 | CD40L, G-CSF (CSF-3), IFN alpha, IL-15, IL-1RA, IL-7 |
Cat. No. | Analyte | Chim- panzee | Afr. gr. monkey | Ba-boon | Cyno- molgus | Pigtail macaque | Rhesus monkey | Mar- moset | Squirrel monkey |
---|---|---|---|---|---|---|---|---|---|
EPX01A-42032-901 | Adiponectin | +++ | N.T. | + | + | N.T. | ++ | - | - |
EPX01A-42116-901 | BDNF | +++ | ++ | + | + | + | ++ | + | + |
EPX01A-42147-901 | BLC/CXCL13 | +++ | +++ | +++ | +++ | ++ | +++ | ++ | + |
EPX01A-40239-901 | CD40L | +++ | + | +++ | - | - | +++ | +++ | + |
EPX01A-42120-901 | Eotaxin | +++ | + | +++ | +++ | +++ | +++ | +++ | ++ |
EPX01A-42074-901 | FGF-2 | +++ | +++ | ++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-42001-901 | G-CSF | +++ | + | +++ | +++ | +++ | +++ | ++ | + |
EPX01A-40283-901 | GM-CSF | +++ | +++ | + | +++ | + | + | + | + |
EPX01A-42027-901 | Granzyme B | +++ | - | - | - | - | - | - | - |
EPX01A-40201-901 | ICAM-1 | +++ | +++ | +++ | +++ | +++ | +++ | ++ | ++ |
EPX01A-40216-901 | IFN alpha | ++ | - | + | ++ | - | ++ | ++ | - |
EPX01A-40228-901 | IFN gamma | +++ | +++ | +++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-40224-901 | IL-1 beta | +++ | + | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-40642-901 | IL-10 | +++ | + | + | + | - | + | ++ | + |
EPX01A-42090-901 | IL-12p40 | +++ | +++ | +++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-40238-901 | IL-12p70 | +++ | ++ | ++ | + | + | + | +++ | +++ |
EPX01A-40231-901 | IL-13 | +++ | + | +++ | + | +/- | ++ | +/- | +/- |
EPX01A-42089-901 | IL-15 | +++ | ++ | ++ | ++ | + | +++ | +++ | + |
EPX01A-42017-901 | IL-17A | +++ | +++ | ++ | ++ | ++ | +++ | +++ | + |
EPX01A-42160-901 | IL-17F | +++ | ++ | ++ | ++ | ++ | ++ | + | + |
EPX01A-40267-901 | IL-18 | +++ | +++ | ++ | ++ | ++ | ++ | ++ | + |
EPX01A-42080-901 | IL-1RA | +++ | ++ | +++ | +++ | +++ | +++ | + | +/- |
EPX01A-40221-901 | IL-2 | +++ | - | - | - | - | - | + | + |
EPX01A-42023-901 | IL-23 | +++ | ++ | ++ | ++ | ++ | +++ | ++ | ++ |
EPX01A-40225-901 | IL-4 | +++ | +++ | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-40278-901 | IL-5 | +++ | ++ | +++ | ++ | + | ++ | ++ | - |
EPX01A-40213-901 | IL-6 | +++ | + | ++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-40237-901 | IL-7 | +++ | - | ++ | ++ | - | +++ | +++ | + |
EPX01A-40204-901 | IL-8 | +++ | ++ | +++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-40284-901 | IP-10 | +++ | +/- | +/- | +/- | +/- | +/- | +/- | +/- |
EPX01A-42124-901 | I-TAC | +++ | +++ | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01B-40281-901 | MCP-1 | +++ | +++ | +++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-42125-901 | MCP-3 | + | + | + | + | + | + | +++ | + |
EPX01A-42127-901 | MIF | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-40285-901 | MIG | +++ | + | ++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-42029-901 | MIP-1 alpha | +++ | +++ | +++ | +++ | ++ | +++ | ++ | +++ |
EPX01A-42030-901 | MIP-1 beta | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-42117-901 | NGF beta | +++ | + | ++ | - | - | +++ | +++ | +++ |
EPX01A-42033-901 | PAI-1 | +++ | ++ | + | + | + | +++ | +++ | ++ |
EPX01A-42071-901 | PDGF-BB | +++ | ++ | +++ | +++ | +/- | +++ | +++ | +++ |
EPX01A-40287-901 | RANTES | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-42040-901 | Resistin | +++ | +++ | +++ | ++ | +++ | + | +++ | +/- |
EPX01A-42137-901 | SCF | + | N.T. | N.T. | N.T. | N.T. | + | ++ | + |
EPX01A-42138-901 | SDF-1 alpha | +++ | +++ | +++ | +++ | +++ | +++ | + | +/- |
EPX01A-42139-901 | TGF alpha | ++ | + | + | + | + | - | - | + |
EPX01A-40249-901 | TGF beta 1 | +++ | +++ | ++ | ++ | ++ | +++ | +++ | ++ |
EPX01A-40223-901 | TNF alpha | +++ | ++ | +++ | +++ | +++ | +++ | +++ | + |
EPX01A-40202-901 | TNF beta | +++ | ++ | ++ | +++ | ++ | +++ | - | - |
EPX01A-42004-901 | TRAIL | +++ | ++ | +++ | ++ | ++ | ++ | ++ | ++ |
EPX01A-40232-901 | VCAM-1 | +++ | +/- | +/- | - | +/- | - | - | - |
EPX01A-40277-901 | VEGF A | +++ | +++ | +++ | +++ | +++ | +++ | +++ | ++ |
EPX01A-42076-901 | VEGF D | N.T. | N.T. | N.T. | N.T. | N.T. | N.T. | ++ | N.T. |
N.T. = Not tested
All preconfigured multi-analyte reagent panels are extensively tested for combinability, interference, and cross-reactivity and go through stringent quality control standards.
Specification | Description |
---|---|
Specificity | Proof of endogenous, native protein detection in specific samples |
Sensitivity | Detection of relevant native protein levels |
Precision | Inter-assay CV: <15%, Intra-assay CV: <15% |
Linearity of dilution | Linear results over the range of the assay |
Lot-to-lot consistency | Each new batch is calibrated against the reference standard lot |
Dynamic range | Large dynamic range of all ProcartaPlex assays |
Cross-reactivity in % | |||||||||||||||
BTLA | GITR | HVEM | IDO | LAG-3 | PD-1 | PD-L1 | PD-L2 | TIM-3 | CD28 | CD80 | CD137 | CD27 | CD152 | ||
Each individual recombinant protein spiked in S1 concentration. | BTLA | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% |
GITR | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 2,9% | 0,0% | 4,4% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
HVEM | 0.0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
IDO | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
LAG-3 | 0,2% | 1,3% | 0,3% | 0,9% | -- | 0,8% | 1,8% | 2,7% | 9,8% | 0,8% | 0,2% | 0,8% | 0,9% | 1,3% | |
PD-1 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
PD-L1 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
PD-L2 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
TIM-3 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
CD28 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | |
CD80 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | |
CD137 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | |
CD27 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | |
CD152 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- |
Cross-reactivity testing of analytes in ProcartaPlex preconfigured panels.
Human multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Human panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Immune Response 80-Plex Human Panel | EPX800-10080-901 | 80 | APRIL, BAFF, BLC, bNGF, CCL1 (I-309), CCL17 (TARC), CCL21 (6Ckine/SLC), CCL23 (MPIF), CCL25 (TECK), CD30, CD40L, CXCL6 (GCP-2), ENA-78, Eotaxin, Eotaxin-2, Eotaxin-3, FGF-2, Fractalkine, Gal-3, G-CSF, GM-CSF, Granzyme A, Granzyme B, Gro a, HGF, IFN-a, IFN-g, IL-10, IL-12p70, IL-13, IL-15, IL-16, IL-17A, IL-18, IL-1a, IL-1b, IL-2, IL-20, IL-21, IL-22, IL-23, IL-27, IL-2R, IL-3, IL-31, IL-34, IL-37 , IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IP-10, I-TAC, LIF, MCP-1, MCP-2, MCP-3, MCP-4 (CCL13), M-CSF, MDC, MIF, MIG, MIP1a, MIP-1b, MIP-2 alpha (CXCL2), MIP-3 beta (CCL19), MIP-3a, MMP-1, PTX3, SCF, TNF b, TNF-a, TNF-R2, TRAIL, TREM-1, TSLP, TWEAK, VEGF-A |
Immune Monitoring 65-Plex Human Panel | EPX650-10065-901 | 65 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-16, IL-17A (CTLA-8), IL-18, IL-20, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, M-CSF, MIF, TNF alpha, TNF beta, TSLP, BLC (CXCL13), ENA-78 (CXCL5), Eotaxin (CCL11), Eotaxin-2 (CCL24), Eotaxin-3 (CCL26), Fractalkine (CX3CL1), Gro-alpha (CXCL1), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (CCL7), MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-3 alpha (CCL20), SDF-1 alpha (CXCL12), FGF-2, HGF, MMP-1, NGF beta, SCF, VEGF-A, APRIL, BAFF, CD30, CD40L (CD154), IL-2R (CD25), TNF-RII, TRAIL (CD253), TWEAK |
Cytokine/Chemokine/Growth Factor Convenience 45-Plex Human Panel 1 | EPXR450-12171-901 | 45 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, SCF, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, BDNF, EGF, FGF-2, HGF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Immune Checkpoint 37-Plex Human Panel | EPX370-15846-901 | 37 | Arginase-1, B7-H6, BTLA, CD134 (OX40), CD137 (4-1BB), CD152 (CTLA4), CD27, CD276 (B7-H3), CD28, CD47 (IAP), CD48 (BLAST-1), CD73 (NT5E), CD80, CD96 (Tactile), E-cadherin, GITR, HVEM, ICOS ligand (B7-H2), IDO, LAG-3, MICA, MICB, nectin-2, PD-1, PD-L1, PD-L2, perforin, PVR, S100A8/A9, siglec-7, siglec-9, TIM-3, TIMD-4, ULBP-1, ULBP-3, ULBP-4, VISTA (B7-H5) |
Cytokine & Chemokine Convenience 34-Plex Human Panel 1A | EPXR340-12167-901 | 34 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Abundant Serum Markers 26-Plex Human Panel | EPX260-15808-901 | 26 | Adiponectin, CD14, CD44, CD44var (v6), CD62L (L-Selectin), Cystatin C, Elafin, Endoglin, ICAM-1, Lactoferrin, Leptin, MBL, MMP-3, MMP-9, NGAL, NRP-1, Osteopontin (OPN), PAI-1 (Serpin), RANTES (CCL5), RBP4, REG-3a, Resistin (ADSF), SCGF beta, TIMP-1, VCAM-1, ZAG |
Cytokine Storm 21-Plex Human Panel | EPX210-15850-901 | 21 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), TNF alpha, TNF beta |
Inflammation 20-Plex Human Panel | EPX200-12185-901 | 20 | CD62P (P-Selectin), CD62E (E-selectin), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IP-10 (CXCL10), ICAM-1, MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), TNF alpha |
Angiogenesis 18-Plex Human Panel | EPX180-15806-901 | 18 | Angiopoietin-1, BMP-9, CD31 (PECAM-1), EGF, EMMPRIN, Follistatin, FGF-2, G-CSF (CSF-3), HB-EGF, HGF, IL-8 (CXCL8), Leptin, LYVE-1, PDGF-BB, Syndecan, TIE-2, VEGF-A, VEGF-D |
Neuroscience 18-plex Human Panel | EPX180-15837-901 | 18 | Amyloid beta 1-42, BDNF, YKL-40 (CHI3L1), CNTF, FGF-21, GDNF, GFAP, Kallikrein-6 (KLK6), MIF, NCAM-1, Neurogranin (NRGN), NF-H, NGF beta, S100B, Tau (total), Tau (pT181), TDP-43, UCHL1 |
TNF Superfamily 15-Plex Human Panel | EPX150-15801-901 | 15 | CD137L (4-1BBL), APRIL, BAFF, CD30L, CD40L, EDA-1, Fas-L, GITRL, LIGHT (CD258), NGF beta, RANKL, TNF alpha, TNF beta, TRAIL, TWEAK |
Immuno-Oncology Checkpoint 14-Plex Human Panel 1 | EPX14A-15803-901 | 14 | BTLA, CD80, CD152 (CTLA4), CD27, CD28, CD137 (4-1BB), GITR, HVEM, IDO, LAG-3, PD-1, PD-L1, PD-L2, TIM-3 |
Immuno-Oncology Checkpoint 14-Plex Human Panel 2 | EPX140-15815-901 | 14 | Arginase-1, CD73 (NT5E), CD96 (Tactile), E-Cadherin, Nectin-2, MICA, MICB, PVR, Siglec-7, Perforin, Siglec-9, ULBP-1, ULBP-3, ULBP-4 |
Cell Proliferation and Metastasis 13-Plex Human Panel 1 | EPX130-15841-901 | 13 | AFP (alpha-fetoprotein), ADAM12, beta-catenin, CD171 (NCAM-L1), CD66f (PSBG-1), galectin-1, hCG, HO-1 (heme oxygenase 1), IGFBP-1, N-cadherin, nectin-4, PAPP-A, thrombomodulin (TM) 1 |
Apoptotic Cell Clearance 12-Plex Human Panel | EPX120-15816-901 | 12 | AXL, CD36, LOX-1, Mer (MERTK), CD31 (PECAM-1), RAGE, TYRO3, Gas6, MBL ,uPAR, CALR (CRT), PAI-1 (Serpin) |
Cell Proliferation and Metastasis 12-Plex Human Panel 2 | EPX120-15842-901 | 12 | beta-2-microglobulin (B2M), cathepsin D, CEA (CEACAM-5), EGFR (ErbB1), haptoglobin, HGFR (c-Met), IGFBP-2, IGFBP-3, MIA, MIP-4 (CCL18), periostin (OSF-2), VE-cadherin |
Endothelial Injury Marker 12-Plex Human Panel | EPX120-15849-901 | 12 | E-selectin, P-selectin, VEGF-A, VEGF-D, PECAM-1, Angiopoietin-1, Syndecan-1, VEGF-R2, VEGF-R3, MMP-1, Thrombomodulin (TM), VLA-4 |
TNF Receptor Superfamily 12-Plex Human Panel | EPX120-15802-901 | 12 | CD27, CD30, CD40, CD137 (4-1BB), DR6, Fas (APO), GITR, HVEM, TNF-RI, TNF-RII, TRAIL-R1, TRAIL-R2 |
Coronavirus Ig Total Human 11-Plex Panel | EPX110-16000-901 | 11 | SARS-CoV-2 Spike (trimer), SARS-CoV-2 S1 protein, SARS-CoV-2 RBD (IAP), SARS-CoV-2 Nucleocapsid, SARS S1, CoV-NL63, CoV-HKU1, CoV-229E, MERS S1, CoV-OC43, Negative control |
Kidney Toxicity Panel 1 | EPX110-15855-901 | 11 | KIM-1 (TIM-1/HAVCR1), Renin, Calbindin, Osteoactivin (GPNMB), GSTA1 (Glutathione S-transferase A1), IL-18, IP-10 (CXCL10), MCP-1 (CCL2), VEGF-A, Clusterin (APO-J), RBP4 (Retinol-binding protein 4) |
Kidney Toxicity Panel 2 | EPX090-15856-901 | 9 | Uromodulin, Alpha-1-Microglobulin, TFF3 (Trefoil Factor 3), Osteopontin (OPN), Cystatin C, NGAL, Beta-2-microglobulin (B2M), TIMP-1, EGF |
Atherosclerosis 9-Plex Human Panel 1 | EPX090-15822-901 | 9 | FGF-2, HSP60, IL-6, IL-8 (CXCL8), LOX-1, Myeloperoxidase (MPO), PAI-1 (Serpin), Tenascin-C, TRAIL |
Chemokine 9-Plex Human Panel 2 | EPX090-15840-901 | 9 | CCL1 (I-309), CCL17 (TARC), CCL21 (6Ckine/SLC), CCL23 (MPIF), CCL25 (TECK), CXCL6 (GCP-2), MCP-4 (CCL13), MIP-2 alpha (CXCL2), MIP-3 beta (CCL19) |
Immuno-Oncology Checkpoint 9-Plex Human Panel 3 | EPX090-15820-901 | 9 | B7-H6, CD276 (B7-H3), CD47 (IAP), CD48 (BLAST-1), CD134 (OX40), ICOS Ligand (B7-H2), TIMD-4, S100A8/A9, VISTA (B7-H5) |
Neurodegeneration 9-plex Human Panel 1 | EPX090-15836-901 | 9 | Amyloid beta 1-40, Amyloid beta 1-42, FGF-21, Kallikrein-6 (KLK-6), NCAM-1, Neurogranin (NRGN), Tau (total), Tau (pT181), TDP-43 |
Cardiac Remodeling 8-Plex Human Panel | EPX080-15819-901 | 8 | Galectin-3, IL-33, IL-33R (ST2), IP-10 (CXCL10), LAP, Osteopontin (OPN), PTX3, TREM-1 |
Cell Proliferation 8-Plex Human Panel | EPX080-15844-901 | 8 | EpCAM, glypican-1 (GPC1), HSP27, HSP60, IGF-2, midkine (MK), SPARC (osteonectin), TrkB |
Graft Versus Host Disease 8-Plex Human Panel | EPX080-15814-901 | 8 | CD30, Granzyme A, HGF, IL-2R, IL-33R (ST2), KRT18, Syndecan, TNF-RI |
Myokine 8-Plex Human Panel | EPX080-12186-901 | 8 | BDNF, IL-1RA, IL-10, IL-15, IL-8 (CXCL8), IL-6, LIF, TNF alpha |
Antibody Isotyping 7-Plex Human Panel | EPX070-10818-901 | 7 | IgG1, IgG2, IgG3, IgG4, IgA, IgE, IgM |
Cardiac Disease 7-Plex Human Panel | EPX070-15809-901 | 7 | Cardiotrophin-1 (CTF-1), CKMB, FABP-3, GDF-15, myoglobin, NTproBNP, troponin I (TNI) |
Apoptosis 6-Plex Human Panel | EPX06A-15823-901 | 6 | Bcl-2, Caspase-3 (active), cleaved PARP, Cytochrome C, GAPDH, p53 |
Coagulation 6-Plex Human Panel 1 | EPX060-10824-901 | 6 | Antithrombin, CRP, Factor XI, Factor XII, Factor XIII, Prothrombin |
Essential Th1/Th2 Cytokine 6-Plex Human Panel | EPX060-10009-901 | 6 | IFN gamma, IL-12p70, IL-4, IL-5, IL-6, TNF alpha |
Exosome Characterization 6-Plex Human Panel | EPX060-15845-901 | 6 | CD9, CD63, CD81, cytochrome c, syntenin-1, VLA-4 |
SARS-CoV-2 Variants Neutralizing Antibody 6-Plex Panel | EPX060-16018-901 | 6 | SARS-CoV-2 S1 B.1.1.529 (o), SARS-CoV-2 S1 B.1.617.2 (δ), SARS-CoV-2 Spike B.1.617.2 P.1 (γ), SARS-CoV-2 S1 B.1.351 (β), SARS-CoV-2 S1 B.1.1.7 (α), SARS-CoV-2 S1 (wt) |
Apolipoprotein 5-Plex Human Panel | EPX050-15818-901 | 5 | APO A1, APO A2, APO B, APO C2, APO E |
Arteriosclerosis 5-Plex Human Panel | EPX050-15821-901 | 5 | BMP-2, COMP, FGF-23, Osteoprotegerin (OPG), RANKL |
MMP 5-Plex Human Panel 1 | EPX050-10015-901 | 5 | MMP-1, MMP-7, MMP-8, MMP-12, MMP-13 |
Atherosclerosis 4-Plex Human Panel 2 | EPX040-15824-901 | 4 | Lp-PLA2, YKL-40 (CHI3L1), ICAM-1, Adiponectin |
Brain Injury 4-plex Human Panel | EPX040-15827-901 | 4 | NF-H, GFAP, UCHL1, S100B |
Cell Death 4-Plex Human Panel | EPX040-15843-901 | 4 | Dkk-1, HMGB1 (HMG-1), HSP70, HSP90 |
Coagulation 4-Plex Human Panel 3 | EPX040-10825-901 | 4 | Factor IX, Protein C, Protein S, vWF |
Essential 4-Plex Human Panel 2 | EPX040-10008-901 | 4 | IL-1 beta, IL-10, IL-17A (CTLA-8), IL-8 (CXCL8) |
Neurodegeneration 4-plex Human Panel 2 | EPX040-15832-901 | 4 | Apolipoprotein E4, Clusterin (Apo J), Complement Factor H, Fetuin-A |
Neurotrophic Factors 4-plex Human Panel | EPX040-15828-901 | 4 | GDNF, CNTF, BDNF, NGF beta |
Angiogenesis 3-Plex Human Panel 2 | EPX030-15807-901 | 3 | Angiogenin, Angiostatin, Endostatin |
MMP 3-Plex Human Panel 2 | EPX03A-10829-901 | 2 | MMP-2, MMP-3, MMP-9 |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Human Panel Name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 45-Plex Human Panel 1 | EPX450-12171-901 | 45 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, LIF, SCF, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, BDNF, EGF, FGF-2, HGF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Cytokine Chemokine 34-Plex Human Panel 1A | EPX340-12167-901 | 34 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta, Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Cytokine 25-Plex Human Panel 1B | EPX250-12166-901 | 25 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNF alpha, TNF beta |
Th1/Th2 & Chemokine 20-Plex Human Panel 1 | EPX200-12173-901 | 20 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8, IL-12p70, IL-13, IL-18, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, TNF alpha |
Th1/Th2 & Cytokine 18-Plex Human Panel 1C | EPX180-12172-901 | 18 | GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-12p70, IL-13, IL-15, IL-18, IL-31, TNF alpha, TNF beta |
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 18-Plex Human Panel | EPX180-12165-901 | 18 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, TNF alpha |
Th9/Th17/Th22/Treg & Chemokine 16-Plex Human Panel 1 | EPX160-12175-901 | 16 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-8 (CXCL8), IL-9, IL-10, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha |
Cytokine 1C & Chemokine 16-Plex Human Panel 1 | EPX160-12176-901 | 16 | Eotaxin (CCL11), GRO alpha (CXCL1), IFN alpha, IL-1 alpha, IL-1RA, IL-7, IL-8 (CXCL8), IL-15, IL-31, GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), RANTES (CCL5), SDF-1 alpha, TNF beta |
Th9/Th17/Th22/Treg & Cytokine 14-Plex Human Panel 1C | EPX140-12174-901 | 14 | IFN alpha, IL-1 alpha, IL-1RA, IL-7, IL-9, IL-10, IL-15, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IL-31, TNF beta |
Growth Factor 11-Plex Human Panel | EPX110-12170-901 | 11 | BDNF, EGF, FGF-2, HGF, LIF, NGF beta, PDGF-BB, PlGF-1, SCF, VEGF-A, VEGF-D |
Th1/Th2 Cytokine 11-Plex Human Panel | EPX110-10810-90 | 11 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha |
Chemokine 9-Plex Human Panel 1 | EPX090-12187-901 | 9 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-8 (CXCL8), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha, RANTES (CCL5) |
Th9/Th17/Th22/Treg Cytokine 7-Plex Human Panel | EPX070-10817-901 | 7 | IL-10, IL-17A (CTLA-8), IL-21, IL-22, IL-23, IL-27, IL-9 |
Cytokine 7-Plex Human Panel 1C | EPX070-10010-90 | 7 | IFN alpha, IL-31, IL-15, IL-1 alpha, IL-1RA, IL-7, TNF beta |
The ProcartaPlex high-sensitivity immunoassays measure protein targets significantly below the detection limit of conventional assays for Luminex systems. Detect and quantify key cytokines that are expressed in the low pg/mL range.
HS panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
9-Plex Human Panel, High Sensitivity | EPXS090-12199-901 | 9 | IFN gamma, IL-1 beta, IL-2, IL-4, IL-6, IL-10, IL-12p70, IL-17A (CTLA-8), TNF alpha |
Mouse multiplex preconfigured panels are provided in a ready to use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Mouse panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Immune Response 64-Plex Mouse Panel | EPX640-20064-901 | 64 | BAFF, BLC (CXCL13), BTC, CD27, CTACK (CCL27), ENA-78 (CXCL5), Eotaxin (CCL11), Eotaxin-2 (CCL24), G-CSF (CSF-3), GM-CSF, Granzyme B, GRO alpha (CXCL1), IFN alpha, IFN gamma, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-16, IL-17A (CTLA-8), IL-18, IL-19, IL-1 alpha, IL-1 beta, IL-2, IL-22, IL-23, IL-25 (IL-17E), IL-27, IL-28, IL-2Ra, IL-3, IL-31, IL-33, IL-33R (ST2), IL-4, IL-5, IL-6, IL6R/sIL-6R, IL-7, IL-7R alpha, IL-9, IP-10 (CXCL10), I-TAC (CXCL11), Leptin, LIF, MCP-1 (CCL2), MCP-3 (CCL7), MCP-5 (CCL12), M-CSF, MDC (CCL22), MIP-1 alpha, MIP-1 beta, MIP-2, MIP-3b (CCL19), RANKL, RANTES (CCL5), SYB16 (CXCL16), TARC (CCL17), TECK (CCL25), TNF alpha, TSLP, VEGF-A, VEGF-R2 (KDR) |
Immune Monitoring 48-Plex Mouse Panel | EPX480-20834-901 | 48 | BAFF, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-19, IL-22, IL-23, IL-25 (IL-17E), IL-27, IL-28, IL-31, IL-33, LIF, M-CSF, RANKL, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), Betacellulin (BTC), Leptin, VEGF-A, IL-2R, IL-7R alpha, IL-33R (ST2) |
Cytokine & Chemokine Convenience 36-Plex Mouse Panel 1A | EPXR360-26092-901 | 36 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, LIF, M-CSF, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Cytokine & Chemokine Convenience 26-Plex Mouse Panel 1 | EPXR260-26088-901 | 26 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IP-10 (CXCL10), IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), TNF alpha |
Antibody Isotyping 7-Plex Mouse Panel 2 | EPX070-20816-901 | 7 | IgG1, IgG2b, IgG2c, IgG3, IgA, IgE, IgM |
Antibody Isotyping 7-Plex Mouse Panel | EPX070-20815-901 | 7 | IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, IgM |
Immuno-Oncology Checkpoint 7-Plex Mouse Panel 2 | EPX070-20835-901 | 7 | CD137L (4-1BBL), CD152 (CTLA4), CD276 (B7-H3), CD80, PD-1, PD-L1 |
Myokine 5-Plex Mouse Panel | EPX050-20833-901 | 5 | IL-10, IL-15/IL-15R, IL-6, LIF, TNF alpha |
Immuno-Oncology Checkpoint 4-Plex Mouse Panel 1 | EPX040-20830-901 | 4 | TIM-3, LAG-3, BTLA, CD27 |
IFN alpha/IFN beta 2-Plex Mouse Panel | EPX02A-22187-901 | 2 | IFN alpha, IFN beta |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Mouse panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 36-Plex Mouse Panel 1A | EPX360-26092-901 | 36 | G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-15/IL-15R, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, LIF, M-CSF, TNF alpha, ENA-78 (CXCL5), Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Cytokine & Chemokine 26-Plex Mouse Panel 1 | EPX260-26088-901 | 26 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IP-10 (CXCL10), IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5), TNF alpha |
Th1/Th2 & Chemokine 20-Plex Mouse Panel 1 | EPX200-26090-901 | 20 | Eotaxin (CCL11), GRO alpha (CXCL1), GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha, IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 17-Plex Mouse Panel | EPX170-26087-901 | 17 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-22, IL-23, IL-27, TNF alpha |
Th9/Th17/Th22/Treg & Chemokine 15-Plex Mouse Panel | EPX150-26089-901 | 15 | Eotaxin (CCL11), GRO alpha (CXCL1), IL-9, IL-10, IL-17A (CTLA-8), IL-22, IL-23, IL-27, IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Th1/Th2 Cytokine 11-Plex Mouse Panel | EPX110-20820-901 | 11 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-12p70, IL-13, IL-18, TNF alpha |
Cytokine 10-Plex Mouse Panel 1B | EPX100-26091-901 | 10 | ENA-78 (CXCL5), G-CSF (CSF-3), IFN alpha, IL-1 alpha, IL-3, IL-15/IL-15R, IL-28, IL-31, LIF, M-CSF |
Chemokine 9-Plex Mouse Panel 1 | EPX090-20821-901 | 9 | Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2, RANTES (CCL5) |
Essential 8-Plex Mouse Panel 2 | EPX080-20832-901 | 8 | GRO alpha (CXCL1), IL-1 beta, IL-10, IL-17A (CTLA-8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-2 |
Th9/Th17/Th22/Treg Cytokine 6-Plex Mouse Panel | EPX060-20822-901 | 6 | IL-10, IL-17A (CTLA-8), IL-22, IL-23, IL-27, IL-9 |
Essential Th1/Th2 Cytokine 6-Plex Mouse Panel | EPX060-20831-901 | 6 | IFN gamma, IL-12p70, IL-4, IL-5, IL-6, TNF alpha |
The ProcartaPlex high-sensitivity immunoassays measure protein targets significantly below the detection limit of conventional assays for Luminex systems. Detect and quantify key cytokines that are expressed in the low pg/mL range.
HS panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
5-Plex Mouse Panel, High Sensitivity | EPXS050-22199-901 | 5 | IFN gamma, IL-2, IL-4, IL-6, TNF alpha |
Rat multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex Assays.
Rat panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Antibody Isotyping 6-Plex Rat Panel | EPX060-30123-901 | 6 | IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM |
EPX050-30124-901 | 5 | Calbindin, KIM-1, Osteopontin (OPN), TFF3, VEGF-A | |
EPX050-30125-901 | 5 | Albumin, Clusterin (Apo-J), Cystatin, NGAL, TIMP-1 |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Rat panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 22-Plex Rat Panel | EPX220-30122-901 | 22 | Eotaxin (CCL11), GRO alpha (CXCL1), G-CSF (CSF-3), GM-CSF, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A(CTLA-8), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-2, RANTES (CCL5), TNF alpha |
Th Complete 14-Plex Rat Panel | EPX140-30120-901 | 14 | G-CSF (CSF-3), GM-CSF, IFN gamma, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), TNF alpha |
Chemokine 8-Plex Rat Panel | EPX080-30121-901 | 8 | Eotaxin (CCL11), GRO alpha (CXCL1), IP-10 (CXCL10), MCP-1 (CCL2), MCP-3 (CCL7), MIP-1 alpha (CCL3), MIP-2, RANTES (CCL5) |
Canine or Porcine multiplex preconfigured panels are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1x, requiring less pipetting and experimental setup. Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex Assays.
Canine panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 11-Plex Canine Panel 1 | EPX11A-50511-901 | 11 | IFN gamma, IL-2, IL-6, IL-8 (CXCL8), IL-10, IL-12/IL-23p40, MCP-1 (CCL2), NGF beta, SCF, TNF alpha, VEGF-A |
Porcine panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine & Chemokine 9-Plex Porcine Panel 1 | EPX090-60829-901 | 9 | IFN alpha, IFN gamma, IL-1 beta, IL-10, IL-12/IL-23p40, IL-4, IL-6, IL-8 (CXCL8), TNF alpha |
Products can be combined with other combinable panels and simplex kits. Detection antibody (50x) needs to be diluted prior to usage. Assays may contain more than one vial of beads and detection antibody.
Non-human primate panel name | Cat. No. | Plex size | Analytes |
---|---|---|---|
Cytokine/Chemokine/Growth Factor 37-Plex NHP Panel | EPX370-40045-901 | 37 | CD40L, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-23, SCF, TNF alpha, BLC, Eotaxin (CCL11), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha, BDNF, FGF-2, NGF beta, PDGF-BB, SCF, VEGF-A, VEGF-D |
Cytokine & Chemokine 30-Plex NHP Panel | EPX300-40044-901 | 30 | CD40L, G-CSF (CSF-3), GM-CSF, IFN alpha, IFN gamma, IL-1 beta, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-12p70, IL-13, IL-15, IL-17A (CTLA-8), IL-18, IL-23, TNF alpha, BLC (CXCL13), Eotaxin (CCL11), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha |
Th 14-Plex NHP Panel | EPX140-40040-901 | 14 | GM-CSF, IFN gamma, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A (CTLA-8), IL-18, IL-23, TNF alpha |
Chemokine 10-Plex NHP Panel | EPX100-40041-901 | 10 | BLC (CXCL13), Eotaxin (CCL11), IL-8 (CXCL8), IP-10 (CXCL10), I-TAC (CXCL11), MCP-1 (CCL2), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), SDF-1 alpha |
Growth Factor 7-Plex NHP Panel | EPX070-40043-901 | 7 | BDNF, FGF-2, NGF beta, PDGF-BB, SCF, VEGF-A, VEGF-D |
Cytokine 6-Plex NHP Panel 1B | EPX060-40042-901 | 6 | CD40L, G-CSF (CSF-3), IFN alpha, IL-15, IL-1RA, IL-7 |
Cat. No. | Analyte | Chim- panzee | Afr. gr. monkey | Ba-boon | Cyno- molgus | Pigtail macaque | Rhesus monkey | Mar- moset | Squirrel monkey |
---|---|---|---|---|---|---|---|---|---|
EPX01A-42032-901 | Adiponectin | +++ | N.T. | + | + | N.T. | ++ | - | - |
EPX01A-42116-901 | BDNF | +++ | ++ | + | + | + | ++ | + | + |
EPX01A-42147-901 | BLC/CXCL13 | +++ | +++ | +++ | +++ | ++ | +++ | ++ | + |
EPX01A-40239-901 | CD40L | +++ | + | +++ | - | - | +++ | +++ | + |
EPX01A-42120-901 | Eotaxin | +++ | + | +++ | +++ | +++ | +++ | +++ | ++ |
EPX01A-42074-901 | FGF-2 | +++ | +++ | ++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-42001-901 | G-CSF | +++ | + | +++ | +++ | +++ | +++ | ++ | + |
EPX01A-40283-901 | GM-CSF | +++ | +++ | + | +++ | + | + | + | + |
EPX01A-42027-901 | Granzyme B | +++ | - | - | - | - | - | - | - |
EPX01A-40201-901 | ICAM-1 | +++ | +++ | +++ | +++ | +++ | +++ | ++ | ++ |
EPX01A-40216-901 | IFN alpha | ++ | - | + | ++ | - | ++ | ++ | - |
EPX01A-40228-901 | IFN gamma | +++ | +++ | +++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-40224-901 | IL-1 beta | +++ | + | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-40642-901 | IL-10 | +++ | + | + | + | - | + | ++ | + |
EPX01A-42090-901 | IL-12p40 | +++ | +++ | +++ | +++ | ++ | +++ | +++ | +++ |
EPX01A-40238-901 | IL-12p70 | +++ | ++ | ++ | + | + | + | +++ | +++ |
EPX01A-40231-901 | IL-13 | +++ | + | +++ | + | +/- | ++ | +/- | +/- |
EPX01A-42089-901 | IL-15 | +++ | ++ | ++ | ++ | + | +++ | +++ | + |
EPX01A-42017-901 | IL-17A | +++ | +++ | ++ | ++ | ++ | +++ | +++ | + |
EPX01A-42160-901 | IL-17F | +++ | ++ | ++ | ++ | ++ | ++ | + | + |
EPX01A-40267-901 | IL-18 | +++ | +++ | ++ | ++ | ++ | ++ | ++ | + |
EPX01A-42080-901 | IL-1RA | +++ | ++ | +++ | +++ | +++ | +++ | + | +/- |
EPX01A-40221-901 | IL-2 | +++ | - | - | - | - | - | + | + |
EPX01A-42023-901 | IL-23 | +++ | ++ | ++ | ++ | ++ | +++ | ++ | ++ |
EPX01A-40225-901 | IL-4 | +++ | +++ | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-40278-901 | IL-5 | +++ | ++ | +++ | ++ | + | ++ | ++ | - |
EPX01A-40213-901 | IL-6 | +++ | + | ++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-40237-901 | IL-7 | +++ | - | ++ | ++ | - | +++ | +++ | + |
EPX01A-40204-901 | IL-8 | +++ | ++ | +++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-40284-901 | IP-10 | +++ | +/- | +/- | +/- | +/- | +/- | +/- | +/- |
EPX01A-42124-901 | I-TAC | +++ | +++ | +++ | +++ | ++ | +++ | ++ | ++ |
EPX01B-40281-901 | MCP-1 | +++ | +++ | +++ | +++ | +++ | +++ | +/- | +/- |
EPX01A-42125-901 | MCP-3 | + | + | + | + | + | + | +++ | + |
EPX01A-42127-901 | MIF | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-40285-901 | MIG | +++ | + | ++ | +++ | ++ | +++ | ++ | ++ |
EPX01A-42029-901 | MIP-1 alpha | +++ | +++ | +++ | +++ | ++ | +++ | ++ | +++ |
EPX01A-42030-901 | MIP-1 beta | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-42117-901 | NGF beta | +++ | + | ++ | - | - | +++ | +++ | +++ |
EPX01A-42033-901 | PAI-1 | +++ | ++ | + | + | + | +++ | +++ | ++ |
EPX01A-42071-901 | PDGF-BB | +++ | ++ | +++ | +++ | +/- | +++ | +++ | +++ |
EPX01A-40287-901 | RANTES | +++ | +++ | +++ | +++ | +++ | +++ | +++ | +++ |
EPX01A-42040-901 | Resistin | +++ | +++ | +++ | ++ | +++ | + | +++ | +/- |
EPX01A-42137-901 | SCF | + | N.T. | N.T. | N.T. | N.T. | + | ++ | + |
EPX01A-42138-901 | SDF-1 alpha | +++ | +++ | +++ | +++ | +++ | +++ | + | +/- |
EPX01A-42139-901 | TGF alpha | ++ | + | + | + | + | - | - | + |
EPX01A-40249-901 | TGF beta 1 | +++ | +++ | ++ | ++ | ++ | +++ | +++ | ++ |
EPX01A-40223-901 | TNF alpha | +++ | ++ | +++ | +++ | +++ | +++ | +++ | + |
EPX01A-40202-901 | TNF beta | +++ | ++ | ++ | +++ | ++ | +++ | - | - |
EPX01A-42004-901 | TRAIL | +++ | ++ | +++ | ++ | ++ | ++ | ++ | ++ |
EPX01A-40232-901 | VCAM-1 | +++ | +/- | +/- | - | +/- | - | - | - |
EPX01A-40277-901 | VEGF A | +++ | +++ | +++ | +++ | +++ | +++ | +++ | ++ |
EPX01A-42076-901 | VEGF D | N.T. | N.T. | N.T. | N.T. | N.T. | N.T. | ++ | N.T. |
N.T. = Not tested
All preconfigured multi-analyte reagent panels are extensively tested for combinability, interference, and cross-reactivity and go through stringent quality control standards.
Specification | Description |
---|---|
Specificity | Proof of endogenous, native protein detection in specific samples |
Sensitivity | Detection of relevant native protein levels |
Precision | Inter-assay CV: <15%, Intra-assay CV: <15% |
Linearity of dilution | Linear results over the range of the assay |
Lot-to-lot consistency | Each new batch is calibrated against the reference standard lot |
Dynamic range | Large dynamic range of all ProcartaPlex assays |
Cross-reactivity in % | |||||||||||||||
BTLA | GITR | HVEM | IDO | LAG-3 | PD-1 | PD-L1 | PD-L2 | TIM-3 | CD28 | CD80 | CD137 | CD27 | CD152 | ||
Each individual recombinant protein spiked in S1 concentration. | BTLA | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% |
GITR | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 2,9% | 0,0% | 4,4% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
HVEM | 0.0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
IDO | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
LAG-3 | 0,2% | 1,3% | 0,3% | 0,9% | -- | 0,8% | 1,8% | 2,7% | 9,8% | 0,8% | 0,2% | 0,8% | 0,9% | 1,3% | |
PD-1 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
PD-L1 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
PD-L2 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
TIM-3 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | |
CD28 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | 0,0% | |
CD80 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | 0,0% | |
CD137 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | 0,0% | |
CD27 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- | 0,0% | |
CD152 | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | 0,0% | -- |
Cross-reactivity testing of analytes in ProcartaPlex preconfigured panels.
Explore Luminex systems: MAGPIX, Luminex 200, and FLEXMAP3D.
Multiplex up to 80 RNA targets from a single biological sample.
Detect very low levels of proteins using as little as 1–5 μL of sample.
Quantitate single analytes with confidence with highly verified Invitrogen ELISA kits.