Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn06377164_g1
          See other CALM2 GE Assays ›
          Gene Symbol:
          Calm2
          Gene Name:
          calmodulin 2
          Gene Aliases:
          Cam
          Chromosome Location:
          Chr.6: 11064750 - 11080134 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn06377164_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC058485.1 - 720 6921326 95
          FQ211494.1 - 720 95
          FQ211970.1 - 716 95
          FQ212579.1 - 716 95
          FQ212640.1 - 716 95
          FQ212869.1 - 720 95
          FQ213028.1 - 716 95
          FQ213029.1 - 714 95
          FQ213091.1 - 716 95
          FQ213200.1 - 719 95
          FQ213216.1 - 716 95
          FQ213310.1 - 609 95
          FQ213366.1 - 715 95
          FQ213496.1 - 719 95
          FQ213520.1 - 720 95
          FQ213526.1 - 716 95
          FQ213581.1 - 716 95
          FQ213598.1 - 716 95
          FQ213637.1 - 716 95
          FQ213695.1 - 716 95
          FQ213703.1 - 716 95
          FQ213760.1 - 716 95
          FQ213762.1 - 721 95
          FQ213913.1 - 716 95
          FQ213941.1 - 720 95
          FQ213981.1 - 717 95
          FQ213985.1 - 610 95
          FQ213999.1 - 715 95
          FQ214059.1 - 720 95
          FQ214181.1 - 720 95
          FQ214262.1 - 720 95
          FQ214491.1 - 716 95
          FQ214500.1 - 716 95
          FQ214519.1 - 716 95
          FQ214525.1 - 716 95
          FQ214540.1 - 720 95
          FQ215317.1 - 716 95
          FQ215600.1 - 720 95
          FQ216471.1 - 717 95
          FQ218091.1 - 716 95
          FQ219917.1 - 1 207 95
          FQ220250.1 - 716 95
          FQ220698.1 - 720 95
          FQ220779.1 - 716 95
          FQ222184.1 - 719 95
          FQ223357.1 - 720 95
          FQ225272.1 - 716 95
          FQ226994.1 - 720 95
          FQ229228.1 - 716 95
          FQ229580.1 - 720 95
          FQ229619.1 - 716 95
          FQ229905.1 - 715 95
          FQ231022.1 - 720 95
          FQ231702.1 - 720 95
          FQ233337.1 - 719 95
          FQ235266.1 - 717 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0