Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223735_s1
          See other KLRB1A GE Assays ›
          Gene Symbol:
          Klrb1a
          Gene Name:
          killer cell lectin-like receptor subfamily B, member 1A
          Gene Aliases:
          NKR-P1A, Nkrp1a
          Chromosome Location:
          Chr.4: 161949189 - 161966642 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04223735_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klrb1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010964.1 NP_001010964.1 6 950 106
          GenBank mRNA FQ232553.1 - 5 815 106
          M62891.1 - 6 950 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klrb1a

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P598304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44092 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P598304 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P598304 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P598304 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P598304 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701882073 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701092758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9148 - -
          Agilent Rat cDNA Microarray (G4105A) 701092758 - -
          Agilent Rat cDNA Microarray (G4105A) 701882073 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865945 - -
          Affymetrix Rat Gene 1.0 ST Array 10865943 - -
          Affymetrix Rat Gene 1.0 ST Array 10865941 - -
          Affymetrix Rat Gene 1.0 ST Array 10865946 - -
          Affymetrix Rat Gene 1.0 ST Array 10865942 - -
          Affymetrix Rat Gene 1.0 ST Array 10865944 - -
          Affymetrix Rat Gene 1.0 ST Array 10865947 - -
          Affymetrix Rat Gene 1.1 ST Array 10865940 - -
          Rat Genome 230 2.0 Array 1370499_at - -
          Rat Genome 230 2.0 Array 1370499_PM_at - -
          Affymetrix HT Rat Focus Array 1370499_PM_at - -
          Affymetrix HT Rat Focus Array 1370499_at - -
          Affymetrix Rat Clariom S Assay PSR0400027290.rn.1 TC0400003834.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027292.rn.1 TC0400003834.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027291.rn.1 TC0400003834.rn.1 RTA TC
          Rat Expression Set 230 1370499_at - -
          Rat Expression Set 230 1370499_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865940 - -
          Rat Genome U34 Set M62891_g_at - -
          Rat Genome U34 Set M62891_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22204047 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375160 - -
          RatRef-12 Expression BeadChip ILMN_57890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0