Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02758831_s1
          See other LOC102553138 GE Assays ›
          Gene Symbol:
          LOC102553138 Gpr68
          Gene Name:
          ovarian cancer G-protein coupled receptor 1-like
          G protein-coupled receptor 68
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.6: 125316222 - 125318417 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 124874151 - 124903949 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02758831_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102553138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006240453.2 XP_006240515.1 1 1681 79
          Gene Symbol: Gpr68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108049.1 NP_001101519.1 1 1512 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003027.rn.1

          Set Membership:

          Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          positive regulation of insulin secretion
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          negative regulation of monocyte differentiation
          cellular response to pH
          positive regulation of osteoclast development
          G protein-coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10931 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10931 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P239155 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10931 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30732 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891680 - -
          Affymetrix Rat Gene 1.0 ST Array 10891683 - -
          Affymetrix Rat Gene 1.1 ST Array 10891679 - -
          Affymetrix Rat Gene 1.1 ST Array 10891681 - -
          Rat Genome 230 2.0 Array 1382319_at - -
          Rat Genome 230 2.0 Array 1382319_PM_at - -
          Rat Genome 230 2.0 Array 1377544_PM_at - -
          Rat Genome 230 2.0 Array 1377544_at - -
          Affymetrix HT Rat Focus Array 1382319_PM_at - -
          Affymetrix HT Rat Focus Array 1377544_at - -
          Affymetrix HT Rat Focus Array 1377544_PM_at - -
          Affymetrix HT Rat Focus Array 1382319_at - -
          Affymetrix Rat Clariom S Assay PSR0600021356.rn.1 TC0600003027.rn.1 RTA TC
          Rat Expression Set 230 1377544_PM_at - -
          Rat Expression Set 230 1382319_PM_at - -
          Rat Expression Set 230 1377544_at - -
          Rat Expression Set 230 1382319_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891679 - -
          Affymetrix Rat Gene 1.0 ST Array 10891681 - -
          Rat Genome U34 Set rc_AI227816_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20793850 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0