Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02749780_s1
          See other UGT1A7C GE Assays ›
          Gene Symbol:
          Ugt1a7c
          Gene Name:
          UDP glucuronosyltransferase 1 family, polypeptide A7C
          Gene Aliases:
          UDPGT 1-7, Ugt1, Ugt1a7, Ugt1a8
          Chromosome Location:
          Chr.9: 95233957 - 95302822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02749780_s1
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt1a7c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130407.2 NP_569091.2 656 136
          GenBank mRNA AF461738.1 - 664 136
          AY435134.1 - 623 136
          BC078732.1 - 659 136
          U75903.1 - 665 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt1a7c

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          estrogen catabolic process
          flavonoid biosynthetic process
          response to organic cyclic compound
          aromatic compound catabolic process
          cellular response to hormone stimulus
          coumarin catabolic process
          flavonoid glucuronidation
          xenobiotic glucuronidation
          glucuronosyltransferase activity

          MicroArray Probes:

          Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925182 - -
          Affymetrix Rat Gene 1.0 ST Array 10925163 - -
          Affymetrix Rat Gene 1.0 ST Array 10925183 - -
          Affymetrix Rat Gene 1.0 ST Array 10925178 - -
          Affymetrix Rat Gene 1.0 ST Array 10925181 - -
          Affymetrix Rat Gene 1.0 ST Array 10925176 - -
          Affymetrix Rat Gene 1.0 ST Array 10925162 - -
          Affymetrix Rat Gene 1.0 ST Array 10925184 - -
          Affymetrix Rat Gene 1.0 ST Array 10925180 - -
          Affymetrix Rat Gene 1.0 ST Array 10925179 - -
          Affymetrix Rat Gene 1.0 ST Array 10925177 - -
          Affymetrix Rat Gene 1.1 ST Array 10925158 - -
          Rat Genome 230 2.0 Array 1387759_PM_s_at - -
          Rat Genome 230 2.0 Array 1370613_s_at - -
          Rat Genome 230 2.0 Array 1370613_PM_s_at - -
          Rat Genome 230 2.0 Array 1387759_s_at - -
          Affymetrix HT Rat Focus Array 1370613_s_at - -
          Affymetrix HT Rat Focus Array 1370613_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0900009385.rn.1 TC0900001092.rn.1 RTA TC
          Rat Expression Set 230 1387759_s_at - -
          Rat Expression Set 230 1387759_PM_s_at - -
          Rat Expression Set 230 1370613_PM_s_at - -
          Rat Expression Set 230 1370613_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925158 - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set D83796_s_at - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set D38062exon_s_at - -
          Rat Genome U34 Set AF039212mRNA_s_at - -
          Rat Genome U34 Set S56937_s_at - -
          Rat Genome U34 Set J05132_s_at - -
          Rat Genome U34 Set J02612mRNA_s_at - -
          Rat Genome U34 Set D83796_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375198 - -
          RatRef-12 Expression BeadChip ILMN_54507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5f68d85665-v9rd4:80/100.66.130.250:80.
          git-commit: a5c360236fb87e3c46c4dd6f38a1a1c611be0521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0