Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346829_mH
          See other LY49I4 GE Assays ›
          Gene Symbol:
          Ly49i4
          Gene Name:
          Ly49 inhibitory receptor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 164515105 - 164536581 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346829_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly49i4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009495.1 NP_001009495.1 1 - 2 199 130
          XM_008763405.1 XP_008761627.1 2 - 3 314 130
          GenBank mRNA AY649834.1 - 1 - 2 199 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004167.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly49i4

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019908 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019908 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019907 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019908 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019907 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26937 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33818 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4213 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866168 - -
          Affymetrix Rat Gene 1.0 ST Array 10866170 - -
          Affymetrix Rat Gene 1.0 ST Array 10866174 - -
          Affymetrix Rat Gene 1.0 ST Array 10866173 - -
          Affymetrix Rat Gene 1.0 ST Array 10866171 - -
          Affymetrix Rat Gene 1.0 ST Array 10866169 - -
          Affymetrix Rat Gene 1.0 ST Array 10866179 - -
          Affymetrix Rat Gene 1.0 ST Array 10866172 - -
          Affymetrix Rat Gene 1.1 ST Array 10866167 - -
          Affymetrix Rat Clariom S Assay PSR0400030019.rn.1 TC0400004167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030020.rn.1 TC0400004167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030021.rn.1 TC0400004167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400030018.rn.1 TC0400004167.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866167 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20708545 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376803 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0