Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02112997_s1
          See other LGALSL GE Assays ›
          Gene Symbol:
          Lgalsl
          Gene Name:
          lectin, galactoside-binding-like
          Gene Aliases:
          Hspc159, RGD1307414
          Chromosome Location:
          Chr.14: 105047915 - 105055421 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02112997_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lgalsl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134730.1 NP_001128202.1 5 1105 129
          GenBank mRNA BC168193.1 - 5 1105 129
          FQ211679.1 - 5 949 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002327.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P795269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P795269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P795269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P795269 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P737844 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778644 - -
          Affymetrix Rat Gene 1.0 ST Array 10778641 - -
          Affymetrix Rat Gene 1.0 ST Array 10778645 - -
          Affymetrix Rat Gene 1.0 ST Array 10778643 - -
          Affymetrix Rat Gene 1.0 ST Array 10778642 - -
          Affymetrix Rat Gene 1.1 ST Array 10778640 - -
          Rat Genome 230 2.0 Array 1373515_at - -
          Rat Genome 230 2.0 Array 1376867_PM_at - -
          Rat Genome 230 2.0 Array 1373515_PM_at - -
          Rat Genome 230 2.0 Array 1376867_at - -
          Affymetrix HT Rat Focus Array 1373515_at - -
          Affymetrix HT Rat Focus Array 1376867_at - -
          Affymetrix HT Rat Focus Array 1373515_PM_at - -
          Affymetrix HT Rat Focus Array 1376867_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400017276.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017269.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017272.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017267.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017268.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017273.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017266.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017270.rn.1 TC1400002327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400017275.rn.1 TC1400002327.rn.1 RTA TC
          Rat Expression Set 230 1373515_PM_at - -
          Rat Expression Set 230 1376867_at - -
          Rat Expression Set 230 1376867_PM_at - -
          Rat Expression Set 230 1373515_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778640 - -
          Rat Genome U34 Set rc_AI031038_at - -
          Rat Genome U34 Set rc_AI175258_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-v5hlb:80/100.66.134.185:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0