Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195488.1 | NP_001182417.1 | 3 | 3760 | 85 | |
GenBank mRNA | FQ211743.1 | - | 1 | 611 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |