Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01762569_g1
          See other CES2H GE Assays ›
          Gene Symbol:
          Ces2h
          Gene Name:
          carboxylesterase 2H
          Gene Aliases:
          Ces2
          Chromosome Location:
          Chr.19: 37052557 - 37067176 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01762569_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces2h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044258.2 NP_001037723.1 6 - 7 957 72
          NM_001190346.1 NP_001177275.1 6 - 7 957 72
          XM_008772523.1 XP_008770745.1 6 - 7 976 72
          XM_008772524.1 XP_008770746.1 7 - 8 1099 72
          GenBank mRNA BC107806.1 - 6 - 7 927 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000463.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to organic substance
          molecular_function
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P605947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20394 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P605947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P605947 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P605947 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P605947 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72239 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807096 - -
          Affymetrix Rat Gene 1.0 ST Array 10807092 - -
          Affymetrix Rat Gene 1.0 ST Array 10807089 - -
          Affymetrix Rat Gene 1.0 ST Array 10807088 - -
          Affymetrix Rat Gene 1.0 ST Array 10807090 - -
          Affymetrix Rat Gene 1.0 ST Array 10807093 - -
          Affymetrix Rat Gene 1.0 ST Array 10807087 - -
          Affymetrix Rat Gene 1.0 ST Array 10807095 - -
          Affymetrix Rat Gene 1.0 ST Array 10807091 - -
          Affymetrix Rat Gene 1.0 ST Array 10807097 - -
          Affymetrix Rat Gene 1.0 ST Array 10807086 - -
          Affymetrix Rat Gene 1.0 ST Array 10807094 - -
          Affymetrix Rat Gene 1.1 ST Array 10807085 - -
          Affymetrix Rat Clariom S Assay PSR1900003650.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003646.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003645.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003648.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003647.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003643.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003644.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003651.rn.1 TC1900000463.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10807085 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21493532 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-57bdf9945f-4p2c8:80/100.66.134.79:80.
          git-commit: baa7f4fdc4fe37bd11ed6fe4edff1157ae59fd06
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0