Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753515_m1
          See other NUDT15 GE Assays ›
          Gene Symbol:
          Nudt15
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 15
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 55417774 - 55456698 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753515_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106049.1 NP_001099519.1 3 - 4 641 80
          XM_006252294.2 XP_006252356.1 2 - 3 956 80
          XM_008770862.1 XP_008769084.1 3 - 4 744 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002041.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          dGTP catabolic process
          purine deoxyribonucleoside triphosphate catabolic process
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142046 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142046 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142046 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54336 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10785253 - -
          Affymetrix Rat Gene 1.0 ST Array 10785254 - -
          Affymetrix Rat Gene 1.0 ST Array 10785252 - -
          Affymetrix Rat Gene 1.1 ST Array 10785251 - -
          Affymetrix Rat Clariom S Assay PSR1500014133.rn.1 TC1500002041.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10785251 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20980470 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358826 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fbcb5ff59-4c8rf:80/100.66.130.250:80.
          git-commit: f911e8b0a61717f033c2f2c0e0d8be5576cef2f2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0