Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530982_g1
          See other LOC292543 GE Assays ›
          Gene Symbol:
          LOC292543
          Gene Name:
          similar to solute carrier family 7 (cationic amino acid transporter, y+ system), member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 64618350 - 64632132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530982_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC292543
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025639.1 NP_001020810.1 11 - 12 1866 107
          XM_006228022.2 XP_006228084.1 11 - 12 1958 107
          XM_008758754.1 XP_008756976.1 11 - 12 2031 107
          GenBank mRNA BC098062.1 - 11 - 12 1866 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000727.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC292543

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          amino acid transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101164 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67934 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10703792 - -
          Affymetrix Rat Gene 1.0 ST Array 10703788 - -
          Affymetrix Rat Gene 1.0 ST Array 10703790 - -
          Affymetrix Rat Gene 1.0 ST Array 10703797 - -
          Affymetrix Rat Gene 1.0 ST Array 10703794 - -
          Affymetrix Rat Gene 1.0 ST Array 10703798 - -
          Affymetrix Rat Gene 1.0 ST Array 10703789 - -
          Affymetrix Rat Gene 1.0 ST Array 10703799 - -
          Affymetrix Rat Gene 1.0 ST Array 10703796 - -
          Affymetrix Rat Gene 1.0 ST Array 10703791 - -
          Affymetrix Rat Gene 1.0 ST Array 10703795 - -
          Affymetrix Rat Gene 1.0 ST Array 10703793 - -
          Affymetrix Rat Gene 1.1 ST Array 10703787 - -
          Affymetrix Rat Clariom S Assay PSR0100004811.rn.1 TC0100000727.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004812.rn.1 TC0100000727.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10703787 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21952257 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352856 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0