Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01526554_g1
          See other CYP51 GE Assays ›
          Gene Symbol:
          Cyp51
          Gene Name:
          cytochrome P450, family 51
          Gene Aliases:
          Cyp51a1, RATCP14DM
          Chromosome Location:
          Chr.4: 27175564 - 27194018 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01526554_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp51
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012941.2 NP_037073.1 9 - 10 1400 145
          GenBank mRNA AY724494.1 - 9 - 10 1722 145
          BC087033.1 - 9 - 10 1400 145
          D55681.1 - 9 - 10 1453 145
          U17697.1 - 9 - 10 1566 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000286.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp51

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          cholesterol biosynthetic process via 24,25-dihydrolanosterol
          regulation of cholesterol biosynthetic process
          demethylation
          iron ion binding
          sterol 14-demethylase activity
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P604399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116807 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604399 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P604399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168285 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90255 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853472 - -
          Affymetrix Rat Gene 1.0 ST Array 10853475 - -
          Affymetrix Rat Gene 1.0 ST Array 10853473 - -
          Affymetrix Rat Gene 1.0 ST Array 10853474 - -
          Affymetrix Rat Gene 1.0 ST Array 10853476 - -
          Affymetrix Rat Gene 1.1 ST Array 10853471 - -
          Rat Genome 230 2.0 Array 1367979_PM_s_at - -
          Rat Genome 230 2.0 Array 1387020_at - -
          Rat Genome 230 2.0 Array 1387020_PM_at - -
          Rat Genome 230 2.0 Array 1367979_s_at - -
          Affymetrix HT Rat Focus Array 1387020_PM_at - -
          Affymetrix HT Rat Focus Array 1367979_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387020_at - -
          Affymetrix HT Rat Focus Array 1367979_s_at - -
          Affymetrix Rat Clariom S Assay PSR0400002027.rn.1 TC0400000286.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400002028.rn.1 TC0400000286.rn.1 RTA TC
          Rat Expression Set 230 1387020_at - -
          Rat Expression Set 230 1367979_PM_s_at - -
          Rat Expression Set 230 1367979_s_at - -
          Rat Expression Set 230 1387020_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853471 - -
          Rat Genome U34 Set rc_AI230255_at - -
          Rat Genome U34 Set rc_AA997614_s_at - -
          Rat Genome U34 Set rc_AA963449_s_at - -
          Rat Genome U34 Set AB004096_at - -
          Rat Genome U34 Set U17697_s_at - -
          Rat Genome U34 Set rc_AA963449_s_at - -
          Rat Genome U34 Set U17697_s_at - -
          Rat Genome U34 Set AB004096_at - -
          Rat Genome U34 Set rc_AI230255_at - -
          Rat Genome U34 Set rc_AA997614_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21068224 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364918 - -
          RatRef-12 Expression BeadChip ILMN_70260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0