Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515621_g1
          See other ARHGEF19 GE Assays ›
          Gene Symbol:
          Arhgef19
          Gene Name:
          Rho guanine nucleotide exchange factor (GEF) 19
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 159755535 - 159773699 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515621_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108692.1 NP_001102162.1 4 - 5 977 59
          XM_008764270.1 XP_008762492.1 2 - 3 564 59
          XM_008764271.1 XP_008762493.1 2 - 3 524 59
          XM_008764272.1 XP_008762494.1 2 - 3 1803 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001813.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of actin cytoskeleton organization
          wound healing

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P291028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P291028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P291028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P291028 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P291028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873691 - -
          Affymetrix Rat Gene 1.0 ST Array 10873697 - -
          Affymetrix Rat Gene 1.0 ST Array 10873696 - -
          Affymetrix Rat Gene 1.0 ST Array 10873701 - -
          Affymetrix Rat Gene 1.0 ST Array 10873693 - -
          Affymetrix Rat Gene 1.0 ST Array 10873705 - -
          Affymetrix Rat Gene 1.0 ST Array 10873695 - -
          Affymetrix Rat Gene 1.0 ST Array 10873703 - -
          Affymetrix Rat Gene 1.0 ST Array 10873704 - -
          Affymetrix Rat Gene 1.0 ST Array 10873694 - -
          Affymetrix Rat Gene 1.0 ST Array 10873702 - -
          Affymetrix Rat Gene 1.0 ST Array 10873690 - -
          Affymetrix Rat Gene 1.0 ST Array 10873698 - -
          Affymetrix Rat Gene 1.0 ST Array 10873692 - -
          Affymetrix Rat Gene 1.0 ST Array 10873700 - -
          Affymetrix Rat Gene 1.0 ST Array 10873699 - -
          Affymetrix Rat Gene 1.1 ST Array 10873688 - -
          Rat Genome 230 2.0 Array 1394494_at - -
          Rat Genome 230 2.0 Array 1394494_PM_at - -
          Affymetrix HT Rat Focus Array 1394494_PM_at - -
          Affymetrix HT Rat Focus Array 1394494_at - -
          Affymetrix Rat Clariom S Assay PSR0500013691.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013688.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013679.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013693.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013700.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013701.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013689.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013690.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013697.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013698.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013699.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013706.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013677.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013692.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013696.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013704.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013705.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013675.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013684.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013694.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013695.rn.1 TC0500001813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013707.rn.1 TC0500001813.rn.1 RTA TC
          Rat Expression Set 230 1394494_at - -
          Rat Expression Set 230 1394494_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873688 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21715083 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bb9fc6869-mlxj9:80/100.66.134.79:80.
          git-commit: 0e41a6b66f4afbfa2ecf938c7a5b11a23c71f2c6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0