Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497686_m1
          See other SCAMP1 GE Assays ›
          Gene Symbol:
          Scamp1
          Gene Name:
          secretory carrier membrane protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 23858021 - 23941128 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497686_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scamp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100636.1 NP_001094106.1 2 - 3 246 129
          GenBank mRNA CB740599.1 - 2 - 3 300 129
          L22079.1 - 2 - 3 166 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002628.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          endocytosis
          protein transport
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P560952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P560952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P560952 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P560952 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P560952 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820521 - -
          Affymetrix Rat Gene 1.0 ST Array 10820518 - -
          Affymetrix Rat Gene 1.0 ST Array 10820512 - -
          Affymetrix Rat Gene 1.0 ST Array 10820517 - -
          Affymetrix Rat Gene 1.0 ST Array 10820515 - -
          Affymetrix Rat Gene 1.0 ST Array 10820513 - -
          Affymetrix Rat Gene 1.0 ST Array 10820520 - -
          Affymetrix Rat Gene 1.0 ST Array 10820519 - -
          Affymetrix Rat Gene 1.0 ST Array 10820516 - -
          Affymetrix Rat Gene 1.0 ST Array 10820514 - -
          Affymetrix Rat Gene 1.0 ST Array 10820522 - -
          Affymetrix Rat Gene 1.1 ST Array 10820511 - -
          Rat Genome 230 2.0 Array 1388819_at - -
          Rat Genome 230 2.0 Array 1370978_at - -
          Rat Genome 230 2.0 Array 1388819_PM_at - -
          Rat Genome 230 2.0 Array 1370978_PM_at - -
          Affymetrix HT Rat Focus Array 1370978_PM_at - -
          Affymetrix HT Rat Focus Array 1388819_at - -
          Affymetrix HT Rat Focus Array 1370978_at - -
          Affymetrix HT Rat Focus Array 1388819_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200018403.rn.1 TC0200002628.rn.1 RTA TC
          Rat Expression Set 230 1370978_at - -
          Rat Expression Set 230 1388819_at - -
          Rat Expression Set 230 1370978_PM_at - -
          Rat Expression Set 230 1388819_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820511 - -
          Rat Genome U34 Set rc_AI073164_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22109222 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0