Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493243_m1
          See other PIWIL2 GE Assays ›
          Gene Symbol:
          Piwil2
          Gene Name:
          piwi-like RNA-mediated gene silencing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 52037991 - 52116290 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493243_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Piwil2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107276.1 NP_001100746.1 8 - 9 1161 72
          XM_006252271.2 XP_006252333.2 8 - 9 1107 72
          XM_008770828.1 XP_008769050.1 8 - 9 1111 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001990.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA 5'-end processing
          spermatogenesis
          germ-line stem cell population maintenance
          gene silencing by RNA
          piRNA metabolic process
          DNA methylation involved in gamete generation
          positive regulation of translation
          oogenesis
          positive regulation of meiosis I
          mRNA binding
          piRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P654933 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P541259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P654933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P654933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P541259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P541259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P654933 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P654933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10785046 - -
          Affymetrix Rat Gene 1.0 ST Array 10785041 - -
          Affymetrix Rat Gene 1.0 ST Array 10785042 - -
          Affymetrix Rat Gene 1.0 ST Array 10785037 - -
          Affymetrix Rat Gene 1.0 ST Array 10785034 - -
          Affymetrix Rat Gene 1.0 ST Array 10785051 - -
          Affymetrix Rat Gene 1.0 ST Array 10785035 - -
          Affymetrix Rat Gene 1.0 ST Array 10785047 - -
          Affymetrix Rat Gene 1.0 ST Array 10785053 - -
          Affymetrix Rat Gene 1.0 ST Array 10785036 - -
          Affymetrix Rat Gene 1.0 ST Array 10785050 - -
          Affymetrix Rat Gene 1.0 ST Array 10785052 - -
          Affymetrix Rat Gene 1.0 ST Array 10785049 - -
          Affymetrix Rat Gene 1.0 ST Array 10785038 - -
          Affymetrix Rat Gene 1.0 ST Array 10785045 - -
          Affymetrix Rat Gene 1.0 ST Array 10785043 - -
          Affymetrix Rat Gene 1.0 ST Array 10785044 - -
          Affymetrix Rat Gene 1.0 ST Array 10785033 - -
          Affymetrix Rat Gene 1.0 ST Array 10785039 - -
          Affymetrix Rat Gene 1.0 ST Array 10785040 - -
          Affymetrix Rat Gene 1.0 ST Array 10785032 - -
          Affymetrix Rat Gene 1.0 ST Array 10785048 - -
          Affymetrix Rat Gene 1.1 ST Array 10785031 - -
          Rat Genome 230 2.0 Array 1384745_PM_at - -
          Rat Genome 230 2.0 Array 1384745_at - -
          Affymetrix Rat Clariom S Assay PSR1500013697.rn.1 TC1500001990.rn.1 RTA TC
          Rat Expression Set 230 1384745_PM_at - -
          Rat Expression Set 230 1384745_at - -
          Affymetrix Rat Gene 1.0 ST Array 10785031 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21259490 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0