Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460649_m1
          See other ZFP692 GE Assays ›
          Gene Symbol:
          Zfp692
          Gene Name:
          zinc finger protein 692
          Gene Aliases:
          RGD1306740, Znf692
          Chromosome Location:
          Chr.10: 43740732 - 43752476 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460649_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp692
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126272.1 NP_001119744.1 5 - 6 993 94
          XM_006246383.2 XP_006246445.1 10 - 11 1704 94
          XM_006246384.2 XP_006246446.1 10 - 11 1645 94
          XM_006246385.2 XP_006246447.1 10 - 11 1392 94
          XM_006246386.2 XP_006246448.1 10 - 11 1287 94
          GenBank mRNA BC166433.1 - 10 - 11 1278 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000668.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P128929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128929 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128929 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P128929 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P509588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733815 - -
          Affymetrix Rat Gene 1.0 ST Array 10733826 - -
          Affymetrix Rat Gene 1.0 ST Array 10733823 - -
          Affymetrix Rat Gene 1.0 ST Array 10733819 - -
          Affymetrix Rat Gene 1.0 ST Array 10733827 - -
          Affymetrix Rat Gene 1.0 ST Array 10733822 - -
          Affymetrix Rat Gene 1.0 ST Array 10733817 - -
          Affymetrix Rat Gene 1.0 ST Array 10733825 - -
          Affymetrix Rat Gene 1.0 ST Array 10733821 - -
          Affymetrix Rat Gene 1.0 ST Array 10733820 - -
          Affymetrix Rat Gene 1.0 ST Array 10733824 - -
          Affymetrix Rat Gene 1.0 ST Array 10733828 - -
          Affymetrix Rat Gene 1.1 ST Array 10733812 - -
          Rat Genome 230 2.0 Array 1379342_PM_at - -
          Rat Genome 230 2.0 Array 1379342_at - -
          Rat Genome 230 2.0 Array 1390332_PM_at - -
          Rat Genome 230 2.0 Array 1390332_at - -
          Affymetrix HT Rat Focus Array 1379342_PM_at - -
          Affymetrix HT Rat Focus Array 1379342_at - -
          Affymetrix HT Rat Focus Array 1390332_PM_at - -
          Affymetrix HT Rat Focus Array 1390332_at - -
          Affymetrix Rat Clariom S Assay PSR1000005748.rn.1 TC1000000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000005746.rn.1 TC1000000668.rn.1 RTA TC
          Rat Expression Set 230 1379342_PM_at - -
          Rat Expression Set 230 1379342_at - -
          Rat Expression Set 230 1390332_PM_at - -
          Rat Expression Set 230 1390332_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733812 - -
          Rat Genome U34 Set rc_AA892391_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21259317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355617 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0