Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460055_m1
          See other RFFL GE Assays ›
          Gene Symbol:
          Rffl
          Gene Name:
          ring finger and FYVE-like domain containing E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 70157621 - 70220614 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460055_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rffl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004068.1 NP_001004068.1 5 - 6 1135 136
          XM_006246969.2 XP_006247031.1 6 - 7 1293 136
          XM_006246970.2 XP_006247032.1 6 - 7 1290 136
          XM_006246973.2 XP_006247035.1 6 - 7 1530 136
          XM_006246974.2 XP_006247036.1 6 - 7 1120 136
          XM_006246975.2 XP_006247037.2 7 - 8 2101 136
          XM_006246977.2 XP_006247039.1 6 - 7 2009 136
          XM_006246978.2 XP_006247040.1 6 - 7 1527 136
          XM_006246979.2 XP_006247041.1 6 - 7 1116 136
          XM_008767954.1 XP_008766176.1 7 - 8 2098 136
          XM_008767955.1 XP_008766177.1 7 - 8 2283 136
          XM_008767956.1 XP_008766178.1 6 - 7 2018 136
          GenBank mRNA AY157969.1 - 5 - 6 912 136
          BC079216.1 - 5 - 6 1135 136
          FQ210900.1 - 6 - 7 1001 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003413.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rffl

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          intracellular protein transport
          biological_process
          regulation of fibroblast migration
          negative regulation of tumor necrosis factor-mediated signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein K48-linked ubiquitination
          negative regulation of signal transduction by p53 class mediator
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
          protease binding
          p53 binding
          molecular_function
          zinc ion binding
          ligase activity
          protein kinase binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089565 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P008904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008904 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P008904 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089565 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P478711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745565 - -
          Affymetrix Rat Gene 1.0 ST Array 10745562 - -
          Affymetrix Rat Gene 1.0 ST Array 10745560 - -
          Affymetrix Rat Gene 1.0 ST Array 10745566 - -
          Affymetrix Rat Gene 1.0 ST Array 10745564 - -
          Affymetrix Rat Gene 1.0 ST Array 10745559 - -
          Affymetrix Rat Gene 1.1 ST Array 10745558 - -
          Rat Genome 230 2.0 Array 1377590_PM_at - -
          Rat Genome 230 2.0 Array 1385504_PM_at - -
          Rat Genome 230 2.0 Array 1377590_at - -
          Rat Genome 230 2.0 Array 1385504_at - -
          Affymetrix HT Rat Focus Array 1377590_PM_at - -
          Affymetrix HT Rat Focus Array 1385504_PM_at - -
          Affymetrix HT Rat Focus Array 1377590_at - -
          Affymetrix HT Rat Focus Array 1385504_at - -
          Affymetrix Rat Clariom S Assay PSR1000030400.rn.1 TC1000003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000030401.rn.1 TC1000003413.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000030397.rn.1 TC1000003413.rn.1 RTA TC
          Rat Expression Set 230 1377590_PM_at - -
          Rat Expression Set 230 1385504_at - -
          Rat Expression Set 230 1385504_PM_at - -
          Rat Expression Set 230 1377590_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745558 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21668681 - -
          ABI Rat Genome Survey Microarray 21879645 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367792 - -
          RatRef-12 Expression BeadChip ILMN_52853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0