Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01457754_g1
          See other LOC688869 GE Assays ›
          Gene Symbol:
          LOC688869
          Gene Name:
          similar to cytochrome c oxidase, subunit VIb polypeptide 1
          Gene Aliases:
          Cox6b1
          Chromosome Location:
          Chr.1: 89075987 - 89084834 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01457754_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC688869
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145273.1 NP_001138745.1 1 - 2 119 74
          XM_006228795.1 XP_006228857.1 2 - 3 227 74
          GenBank mRNA FQ217670.1 - 2 - 3 141 74
          FQ217813.1 - 2 - 3 137 74
          FQ218018.1 - 2 - 3 189 74
          FQ221178.1 - 2 - 3 185 74
          FQ222745.1 - 2 - 3 169 74
          FQ222968.1 - 2 - 3 187 74
          FQ223479.1 - 2 - 3 179 74
          FQ223722.1 - 2 - 3 147 74
          FQ223947.1 - 2 - 3 147 74
          FQ224063.1 - 2 - 3 192 74
          FQ224135.1 - 2 - 3 147 74
          FQ226525.1 - 2 - 3 180 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005160.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          substantia nigra development
          proton transmembrane transport
          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157889 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007073 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078982 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078982 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157889 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007073 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720722 - -
          Affymetrix Rat Gene 1.0 ST Array 10720725 - -
          Affymetrix Rat Gene 1.0 ST Array 10818377 - -
          Affymetrix Rat Gene 1.0 ST Array 10720724 - -
          Affymetrix Rat Gene 1.0 ST Array 10720723 - -
          Affymetrix Rat Gene 1.1 ST Array 10720721 - -
          Affymetrix Rat Gene 1.1 ST Array 10818376 - -
          Affymetrix HT Rat Focus Array 1371321_at - -
          Affymetrix HT Rat Focus Array 1371321_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100041505.rn.1 TC0100005160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041506.rn.1 TC0100005160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041502.rn.1 TC0100005160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041503.rn.1 TC0100005160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041500.rn.1 TC0100005160.rn.1 RTA TC
          Rat Genome U34 Set rc_AI104035_s_at - -
          Rat Genome U34 Set rc_AA946040_at - -
          Rat Genome U34 Set rc_AA946040_at - -
          Rat Genome U34 Set rc_AI104035_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421820 - -
          ABI Rat Genome Survey Microarray 22132724 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362270 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-55c5c7589b-pgs97:80/100.66.130.250:80.
          git-commit: fefcbad4eb9baa7f43a2c44c49530f643471c973
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0