Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444461_mH
          See other LOC100910040 GE Assays ›
          Gene Symbol:
          LOC100910040 Ces2c
          Gene Name:
          liver carboxylesterase-like
          carboxylesterase 2C
          Gene Aliases:
          -
          CESRL4, Ces2, Ces2l
          Chromosome Location:
          Chr.1: 282640678 - 282648353 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 282671420 - 282691073 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01444461_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910040
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749155.3 XP_003749203.1 2 - 3 375 79
          Gene Symbol: Ces2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133586.2 NP_598270.1 310 79
          XM_008760588.1 XP_008758810.1 3 - 4 279 79
          XM_008760589.1 XP_008758811.1 2 - 3 285 79
          GenBank mRNA AB010635.1 - 310 79
          AB191005.1 - 347 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003925.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100910040
          Click here to find siRNAs for Gene Ces2c

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          carboxylic ester hydrolase activity
          acylcarnitine hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P102588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102588 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102588 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P102588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716519 - -
          Affymetrix Rat Gene 1.0 ST Array 10716517 - -
          Affymetrix Rat Gene 1.0 ST Array 10716524 - -
          Affymetrix Rat Gene 1.0 ST Array 10716514 - -
          Affymetrix Rat Gene 1.0 ST Array 10716513 - -
          Affymetrix Rat Gene 1.0 ST Array 10716525 - -
          Affymetrix Rat Gene 1.0 ST Array 10716521 - -
          Affymetrix Rat Gene 1.0 ST Array 10716515 - -
          Affymetrix Rat Gene 1.0 ST Array 10716520 - -
          Affymetrix Rat Gene 1.0 ST Array 10716523 - -
          Affymetrix Rat Gene 1.0 ST Array 10716518 - -
          Affymetrix Rat Gene 1.0 ST Array 10716512 - -
          Affymetrix Rat Gene 1.0 ST Array 10716522 - -
          Affymetrix Rat Gene 1.0 ST Array 10716516 - -
          Affymetrix Rat Gene 1.1 ST Array 10716509 - -
          Rat Genome 230 2.0 Array 1368905_at - -
          Rat Genome 230 2.0 Array 1368905_PM_at - -
          Affymetrix HT Rat Focus Array 1368905_PM_at - -
          Affymetrix HT Rat Focus Array 1368905_at - -
          Affymetrix Rat Clariom S Assay PSR0100032044.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032048.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032053.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032050.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032045.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032049.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032052.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032054.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032046.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032051.rn.1 TC0100003925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032047.rn.1 TC0100003925.rn.1 RTA TC
          Rat Expression Set 230 1368905_PM_at - -
          Rat Expression Set 230 1368905_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716509 - -
          Rat Genome U34 Set AB010635_s_at - -
          Rat Genome U34 Set AB010635_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336800 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357047 - -
          RatRef-12 Expression BeadChip ILMN_60214 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fbcb5ff59-4c8rf:80/100.66.130.250:80.
          git-commit: f911e8b0a61717f033c2f2c0e0d8be5576cef2f2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0