Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442853_m1
          See other TGS1 GE Assays ›
          Gene Symbol:
          Tgs1
          Gene Name:
          trimethylguanosine synthase 1
          Gene Aliases:
          Ncoa6ip
          Chromosome Location:
          Chr.5: 16413356 - 16446881 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442853_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107904.1 NP_001101374.1 9 - 10 2138 78
          XM_006237833.2 XP_006237895.1 9 - 10 2120 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000128.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          7-methylguanosine RNA capping
          7-methylguanosine cap hypermethylation
          RNA trimethylguanosine synthase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P808072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P808072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P808072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867455 - -
          Affymetrix Rat Gene 1.0 ST Array 10867458 - -
          Affymetrix Rat Gene 1.0 ST Array 10867453 - -
          Affymetrix Rat Gene 1.0 ST Array 10867449 - -
          Affymetrix Rat Gene 1.0 ST Array 10867456 - -
          Affymetrix Rat Gene 1.0 ST Array 10867450 - -
          Affymetrix Rat Gene 1.0 ST Array 10867460 - -
          Affymetrix Rat Gene 1.0 ST Array 10867457 - -
          Affymetrix Rat Gene 1.0 ST Array 10867451 - -
          Affymetrix Rat Gene 1.0 ST Array 10867452 - -
          Affymetrix Rat Gene 1.0 ST Array 10867459 - -
          Affymetrix Rat Gene 1.0 ST Array 10867454 - -
          Affymetrix Rat Gene 1.0 ST Array 10867448 - -
          Affymetrix Rat Gene 1.1 ST Array 10867447 - -
          Rat Genome 230 2.0 Array 1393391_PM_at - -
          Rat Genome 230 2.0 Array 1393391_at - -
          Affymetrix Rat Clariom S Assay PSR0500000864.rn.1 TC0500000128.rn.1 RTA TC
          Rat Expression Set 230 1393391_PM_at - -
          Rat Expression Set 230 1393391_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867447 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22231143 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0