Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438987_m1
          See other LAMC3 GE Assays ›
          Gene Symbol:
          Lamc3
          Gene Name:
          laminin gamma 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 9265407 - 9327130 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438987_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107830.1 NP_001101300.1 20 - 21 3579 60
          XM_008761668.1 XP_008759890.1 20 - 21 3677 60
          XM_008761669.1 XP_008759891.1 20 - 21 3701 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005095.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          cell morphogenesis involved in differentiation
          visual perception
          astrocyte development
          retina development in camera-type eye

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122634 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P156846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12999 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P156846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122634 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440971 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82241 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835424 - -
          Affymetrix Rat Gene 1.0 ST Array 10835440 - -
          Affymetrix Rat Gene 1.0 ST Array 10835442 - -
          Affymetrix Rat Gene 1.0 ST Array 10835422 - -
          Affymetrix Rat Gene 1.0 ST Array 10835430 - -
          Affymetrix Rat Gene 1.0 ST Array 10835428 - -
          Affymetrix Rat Gene 1.0 ST Array 10835443 - -
          Affymetrix Rat Gene 1.0 ST Array 10835450 - -
          Affymetrix Rat Gene 1.0 ST Array 10835434 - -
          Affymetrix Rat Gene 1.0 ST Array 10835432 - -
          Affymetrix Rat Gene 1.0 ST Array 10835429 - -
          Affymetrix Rat Gene 1.0 ST Array 10835441 - -
          Affymetrix Rat Gene 1.0 ST Array 10835448 - -
          Affymetrix Rat Gene 1.0 ST Array 10835445 - -
          Affymetrix Rat Gene 1.0 ST Array 10835444 - -
          Affymetrix Rat Gene 1.0 ST Array 10835438 - -
          Affymetrix Rat Gene 1.0 ST Array 10835451 - -
          Affymetrix Rat Gene 1.0 ST Array 10835435 - -
          Affymetrix Rat Gene 1.0 ST Array 10835425 - -
          Affymetrix Rat Gene 1.0 ST Array 10835447 - -
          Affymetrix Rat Gene 1.0 ST Array 10835433 - -
          Affymetrix Rat Gene 1.0 ST Array 10835436 - -
          Affymetrix Rat Gene 1.0 ST Array 10835431 - -
          Affymetrix Rat Gene 1.0 ST Array 10835446 - -
          Affymetrix Rat Gene 1.0 ST Array 10835427 - -
          Affymetrix Rat Gene 1.0 ST Array 10835421 - -
          Affymetrix Rat Gene 1.0 ST Array 10835423 - -
          Affymetrix Rat Gene 1.0 ST Array 10835439 - -
          Affymetrix Rat Gene 1.0 ST Array 10835426 - -
          Affymetrix Rat Gene 1.0 ST Array 10835437 - -
          Affymetrix Rat Gene 1.1 ST Array 10835449 - -
          Affymetrix Rat Gene 1.1 ST Array 10835420 - -
          Rat Genome 230 2.0 Array 1393896_at - -
          Rat Genome 230 2.0 Array 1393896_PM_at - -
          Rat Genome 230 2.0 Array 1397972_at - -
          Rat Genome 230 2.0 Array 1397972_PM_at - -
          Affymetrix HT Rat Focus Array 1393896_at - -
          Affymetrix HT Rat Focus Array 1393896_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300039427.rn.1 TC0300005095.rn.1 RTA TC
          Rat Expression Set 230 1393896_at - -
          Rat Expression Set 230 1393896_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835420 - -
          Affymetrix Rat Gene 1.0 ST Array 10835449 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fbcb5ff59-4c8rf:80/100.66.130.250:80.
          git-commit: f911e8b0a61717f033c2f2c0e0d8be5576cef2f2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0