Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438759_m1
          See other GBGT1 GE Assays ›
          Gene Symbol:
          Gbgt1
          Gene Name:
          globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
          Gene Aliases:
          Abo, Abol1
          Chromosome Location:
          Chr.3: 5109880 - 5126901 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438759_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gbgt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023094.3 NP_075582.3 4 - 5 229 111
          GenBank mRNA AF264018.2 - 4 - 5 229 111
          AF469945.1 - 4 - 5 229 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000123.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gbgt1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          defense response to bacterium
          glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase activity
          fucosylgalactoside 3-alpha-galactosyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P162361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P162361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P162361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P355907 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700523038 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22746 - -
          Agilent Rat cDNA Microarray (G4105A) 700523038 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834575 - -
          Affymetrix Rat Gene 1.0 ST Array 10834571 - -
          Affymetrix Rat Gene 1.0 ST Array 10834569 - -
          Affymetrix Rat Gene 1.0 ST Array 10834570 - -
          Affymetrix Rat Gene 1.0 ST Array 10834572 - -
          Affymetrix Rat Gene 1.0 ST Array 10834587 - -
          Affymetrix Rat Gene 1.0 ST Array 10834574 - -
          Affymetrix Rat Gene 1.0 ST Array 10834573 - -
          Affymetrix Rat Gene 1.1 ST Array 10834567 - -
          Affymetrix Rat Gene 1.1 ST Array 10834584 - -
          Rat Genome 230 2.0 Array 1368798_PM_at - -
          Rat Genome 230 2.0 Array 1387335_PM_s_at - -
          Rat Genome 230 2.0 Array 1393031_PM_at - -
          Rat Genome 230 2.0 Array 1387335_s_at - -
          Rat Genome 230 2.0 Array 1377335_at - -
          Rat Genome 230 2.0 Array 1393031_at - -
          Rat Genome 230 2.0 Array 1368798_at - -
          Rat Genome 230 2.0 Array 1377335_PM_at - -
          Rat Genome 230 2.0 Array 1370498_at - -
          Rat Genome 230 2.0 Array 1370498_PM_at - -
          Affymetrix HT Rat Focus Array 1377335_at - -
          Affymetrix HT Rat Focus Array 1370498_PM_at - -
          Affymetrix HT Rat Focus Array 1387335_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370498_at - -
          Affymetrix HT Rat Focus Array 1387335_s_at - -
          Affymetrix HT Rat Focus Array 1368798_at - -
          Affymetrix HT Rat Focus Array 1377335_PM_at - -
          Affymetrix HT Rat Focus Array 1368798_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300001111.rn.1 TC0300000123.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300001112.rn.1 TC0300000123.rn.1 RTA TC
          Rat Expression Set 230 1387335_PM_s_at - -
          Rat Expression Set 230 1370498_at - -
          Rat Expression Set 230 1370498_PM_at - -
          Rat Expression Set 230 1377335_at - -
          Rat Expression Set 230 1377335_PM_at - -
          Rat Expression Set 230 1393031_PM_at - -
          Rat Expression Set 230 1368798_PM_at - -
          Rat Expression Set 230 1387335_s_at - -
          Rat Expression Set 230 1368798_at - -
          Rat Expression Set 230 1393031_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834567 - -
          Rat Genome U34 Set rc_AA891214_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21276022 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375823 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0