Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437060_m1
          See other PAX5 GE Assays ›
          Gene Symbol:
          Pax5
          Gene Name:
          paired box 5
          Gene Aliases:
          RGD1565236
          Chromosome Location:
          Chr.5: 60007587 - 60191973 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437060_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pax5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109261.1 NP_001102731.1 1 - 2 78 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002651.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          skeletal muscle cell differentiation
          positive regulation of transcription by RNA polymerase II
          negative regulation of histone H3-K9 methylation
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165113 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064099 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064099 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86296 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876498 - -
          Affymetrix Rat Gene 1.0 ST Array 10876500 - -
          Affymetrix Rat Gene 1.1 ST Array 10876497 - -
          Affymetrix Rat Clariom S Assay PSR0500019756.rn.1 TC0500002651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019755.rn.1 TC0500002651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019754.rn.1 TC0500002651.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20889259 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372098 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0