Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436234_g1
          See other ABL1 GE Assays ›
          Gene Symbol:
          Abl1 LOC100909750
          Gene Name:
          ABL proto-oncogene 1, non-receptor tyrosine kinase
          tyrosine-protein kinase ABL1-like
          Gene Aliases:
          Abl
          -
          Chromosome Location:
          Chr.3: 10041820 - 10145076 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 9409198 - 9448279 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01436234_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100850.1 NP_001094320.1 5 - 6 2217 74
          XM_006233918.2 XP_006233980.1 5 - 6 1014 74
          GenBank mRNA DQ294401.1 - 5 - 6 959 74
          DQ294402.1 - 5 - 6 959 74
          Gene Symbol: LOC100909750
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749439.3 XP_003749487.1 1014 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          B-1 B cell homeostasis
          B cell proliferation involved in immune response
          transitional one stage B cell differentiation
          phagocytosis
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          response to oxidative stress
          epidermal growth factor receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          learning or memory
          associative learning
          response to xenobiotic stimulus
          cell migration
          peptidyl-tyrosine phosphorylation
          cerebellum morphogenesis
          negative regulation of cell-cell adhesion
          microspike assembly
          actin cytoskeleton organization
          actin filament polymerization
          negative regulation of BMP signaling pathway
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          regulation of actin cytoskeleton organization
          positive regulation of osteoblast proliferation
          substrate adhesion-dependent cell spreading
          cellular response to oxidative stress
          response to endoplasmic reticulum stress
          peptidyl-tyrosine autophosphorylation
          B cell proliferation
          signal transduction in response to DNA damage
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of neuron apoptotic process
          innate immune response
          positive regulation of vasoconstriction
          negative regulation of mitotic cell cycle
          positive regulation of mitotic cell cycle
          alpha-beta T cell differentiation
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          positive regulation of fibroblast proliferation
          spleen development
          thymus development
          collateral sprouting
          positive regulation of peptidyl-tyrosine phosphorylation
          activated T cell proliferation
          B cell receptor signaling pathway
          neuromuscular process controlling balance
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of oxidoreductase activity
          negative regulation of ubiquitin-protein transferase activity
          regulation of cell cycle
          Bergmann glial cell differentiation
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to transforming growth factor beta stimulus
          response to epinephrine
          negative regulation of protein serine/threonine kinase activity
          actin filament branching
          positive regulation of dendrite development
          negative regulation of phospholipase C activity
          positive regulation of neuron death
          positive regulation of hydrogen peroxide-mediated programmed cell death
          regulation of extracellular matrix organization
          positive regulation of extracellular matrix organization
          glomerular visceral epithelial cell apoptotic process
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          negative regulation of endothelial cell apoptotic process
          regulation of cellular senescence
          negative regulation of cellular senescence
          regulation of response to DNA damage stimulus
          cell differentiation
          regulation of cell population proliferation
          regulation of apoptotic process
          magnesium ion binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          SH3 domain binding
          protein kinase binding
          protein domain specific binding
          syntaxin binding
          manganese ion binding
          actin filament binding
          mitogen-activated protein kinase binding
          proline-rich region binding
          delta-catenin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0300021782.rn.1 TC0300002849.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021783.rn.1 TC0300002849.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0