Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01431898_m1
          See other NUDT7 GE Assays ›
          Gene Symbol:
          Nudt7 LOC102555567
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 7
          peroxisomal coenzyme A diphosphatase NUDT7-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.19: 46455766 - 46470074 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 46455685 - 46470070 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01431898_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108450.1 NP_001101920.1 4 - 5 2213 64
          Gene Symbol: LOC102555567
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006255677.2 XP_006255739.2 4 - 5 574 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000639.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleoside diphosphate metabolic process
          coenzyme A catabolic process
          acetyl-CoA catabolic process
          brown fat cell differentiation
          magnesium ion binding
          acetyl-CoA hydrolase activity
          signaling receptor binding
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          manganese ion binding
          snoRNA binding
          m7G(5')pppN diphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P491393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43815 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491393 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491393 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P491393 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808166 - -
          Affymetrix Rat Gene 1.0 ST Array 10808161 - -
          Affymetrix Rat Gene 1.0 ST Array 10808162 - -
          Affymetrix Rat Gene 1.0 ST Array 10808165 - -
          Affymetrix Rat Gene 1.0 ST Array 10808163 - -
          Affymetrix Rat Gene 1.0 ST Array 10808160 - -
          Affymetrix Rat Gene 1.1 ST Array 10808159 - -
          Rat Genome 230 2.0 Array 1389251_PM_at - -
          Rat Genome 230 2.0 Array 1389251_at - -
          Affymetrix HT Rat Focus Array 1389251_at - -
          Affymetrix HT Rat Focus Array 1389251_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900005443.rn.1 TC1900000639.rn.1 RTA TC
          Rat Expression Set 230 1389251_at - -
          Rat Expression Set 230 1389251_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808159 - -
          Rat Genome U34 Set rc_AA944380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21576442 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0