Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426481_m1
          See other ABCB10 GE Assays ›
          Gene Symbol:
          Abcb10
          Gene Name:
          ATP-binding cassette, subfamily B (MDR/TAP), member 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56742809 - 56772904 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426481_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcb10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012166.1 NP_001012166.1 2 - 3 679 58
          GenBank mRNA BC089900.1 - 2 - 3 679 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001672.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcb10

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metabolic process
          transmembrane transport
          molecular_function
          ATP binding
          ATPase-coupled transmembrane transporter activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126068 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811866 - -
          Affymetrix Rat Gene 1.0 ST Array 10811873 - -
          Affymetrix Rat Gene 1.0 ST Array 10811864 - -
          Affymetrix Rat Gene 1.0 ST Array 10811862 - -
          Affymetrix Rat Gene 1.0 ST Array 10811865 - -
          Affymetrix Rat Gene 1.0 ST Array 10811870 - -
          Affymetrix Rat Gene 1.0 ST Array 10811869 - -
          Affymetrix Rat Gene 1.0 ST Array 10811867 - -
          Affymetrix Rat Gene 1.0 ST Array 10811868 - -
          Affymetrix Rat Gene 1.0 ST Array 10811863 - -
          Affymetrix Rat Gene 1.0 ST Array 10811874 - -
          Affymetrix Rat Gene 1.0 ST Array 10811872 - -
          Affymetrix Rat Gene 1.0 ST Array 10811871 - -
          Affymetrix Rat Gene 1.1 ST Array 10811861 - -
          Rat Genome 230 2.0 Array 1382963_PM_at - -
          Rat Genome 230 2.0 Array 1390781_PM_at - -
          Rat Genome 230 2.0 Array 1390781_at - -
          Rat Genome 230 2.0 Array 1382963_at - -
          Affymetrix HT Rat Focus Array 1390781_at - -
          Affymetrix HT Rat Focus Array 1390781_PM_at - -
          Affymetrix HT Rat Focus Array 1382963_at - -
          Affymetrix HT Rat Focus Array 1382963_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900013508.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013511.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013512.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013513.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013514.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013519.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013518.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013520.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013516.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013509.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013510.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013515.rn.1 TC1900001672.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013517.rn.1 TC1900001672.rn.1 RTA TC
          Rat Expression Set 230 1390781_at - -
          Rat Expression Set 230 1382963_PM_at - -
          Rat Expression Set 230 1382963_at - -
          Rat Expression Set 230 1390781_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811861 - -
          Rat Genome U34 Set rc_AA944536_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22058800 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375315 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0