Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01425427_m1
          See other LOC103690067 GE Assays ›
          Gene Symbol:
          LOC103690067 Tprkb
          Gene Name:
          EKC/KEOPS complex subunit Tprkb
          Tp53rk binding protein
          Gene Aliases:
          -
          Cgi-121, RGD1309786
          Chromosome Location:
          Chr.4: 117772553 - 117787465 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 117589566 - 117604481 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01425427_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103690067
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006236742.2 XP_006236804.1 1211 88
          XM_006236743.2 XP_006236805.1 706 88
          XM_006236745.2 XP_006236807.1 576 88
          XM_006236746.2 XP_006236808.1 490 88
          XM_008763108.1 XP_008761330.1 590 88
          Gene Symbol: Tprkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013926.2 NP_001013948.2 4 - 5 554 88
          XM_003749789.3 XP_003749837.1 4 - 5 505 88
          XM_006236804.2 XP_006236866.1 5 - 6 1211 88
          XM_006236805.2 XP_006236867.1 5 - 6 706 88
          XM_006236808.2 XP_006236870.1 5 - 6 576 88
          GenBank mRNA CK475733.1 - 4 - 5 554 88
          CV112449.1 - 4 - 5 482 88
          FQ234910.1 - 4 - 5 511 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001306.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103690067
          Click here to find siRNAs for Gene Tprkb

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P295361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25717 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P295361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P295361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P295361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P295361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856915 - -
          Affymetrix Rat Gene 1.0 ST Array 10856919 - -
          Affymetrix Rat Gene 1.0 ST Array 10856918 - -
          Affymetrix Rat Gene 1.0 ST Array 10856921 - -
          Affymetrix Rat Gene 1.0 ST Array 10856916 - -
          Affymetrix Rat Gene 1.0 ST Array 10856920 - -
          Affymetrix Rat Gene 1.0 ST Array 10856917 - -
          Affymetrix Rat Gene 1.1 ST Array 10856914 - -
          Rat Genome 230 2.0 Array 1376007_PM_at - -
          Rat Genome 230 2.0 Array 1390972_PM_at - -
          Rat Genome 230 2.0 Array 1376007_at - -
          Rat Genome 230 2.0 Array 1382014_PM_at - -
          Rat Genome 230 2.0 Array 1390972_at - -
          Rat Genome 230 2.0 Array 1382014_at - -
          Affymetrix HT Rat Focus Array 1382014_at - -
          Affymetrix HT Rat Focus Array 1390972_PM_at - -
          Affymetrix HT Rat Focus Array 1390972_at - -
          Affymetrix HT Rat Focus Array 1382014_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400009278.rn.1 TC0400001306.rn.1 RTA TC
          Rat Expression Set 230 1390972_PM_at - -
          Rat Expression Set 230 1382014_at - -
          Rat Expression Set 230 1382014_PM_at - -
          Rat Expression Set 230 1390972_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856914 - -
          Rat Genome U34 Set rc_AA819730_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21179999 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369876 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0