Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421517_m1
          See other APH1B GE Assays ›
          Gene Symbol:
          Aph1b LOC100911293 LOC691658
          Gene Name:
          APH1B gamma secretase subunit
          gamma-secretase subunit APH-1B-like
          similar to anterior pharynx defective 1b homolog
          Gene Aliases:
          APH-1B'
          -
          RGD1559979
          Chromosome Location:
          Chr.8: 72608220 - 72629265 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 72508005 - 72598916 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 72472787 - 72494541 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01421517_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aph1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047090.1 NP_001040555.1 5 - 6 740 92
          GenBank mRNA BC098711.1 - 5 - 6 602 92
          BC166592.1 - 5 - 6 696 92
          DQ672552.1 - 5 - 6 740 92
          Gene Symbol: LOC100911293
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008766401.1 XP_008764623.1 735 92
          Gene Symbol: LOC691658
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_001079175.5 XP_001079175.2 4 - 5 647 92
          XM_003750532.3 XP_003750580.1 5 - 6 769 92
          XM_006243318.2 XP_006243380.1 5 - 6 977 92
          XM_006243319.2 XP_006243381.1 4 - 5 858 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002804.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein processing
          positive regulation of catalytic activity
          proteolysis
          endopeptidase activity
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022477 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010135 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98561 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022477 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022477 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22879 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3743 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918371 - -
          Affymetrix Rat Gene 1.0 ST Array 10918370 - -
          Affymetrix Rat Gene 1.0 ST Array 10918373 - -
          Affymetrix Rat Gene 1.0 ST Array 10918372 - -
          Affymetrix Rat Gene 1.0 ST Array 10918369 - -
          Affymetrix Rat Gene 1.1 ST Array 10918368 - -
          Rat Genome 230 2.0 Array 1388440_at - -
          Rat Genome 230 2.0 Array 1388440_PM_at - -
          Affymetrix HT Rat Focus Array 1388440_PM_at - -
          Affymetrix HT Rat Focus Array 1388440_at - -
          Affymetrix Rat Clariom S Assay PSR0800022799.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022801.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022796.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022798.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022800.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022802.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022803.rn.1 TC0800002804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022794.rn.1 TC0800002804.rn.1 RTA TC
          Rat Expression Set 230 1388440_PM_at - -
          Rat Expression Set 230 1388440_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918368 - -
          Rat Genome U34 Set rc_AI233885_at - -
          Rat Genome U34 Set rc_AI233885_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21779352 - -
          ABI Rat Genome Survey Microarray 22191785 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0