Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418876_g1
          See other RAB12 GE Assays ›
          Gene Symbol:
          Rab12
          Gene Name:
          RAB12, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 114683985 - 114709613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418876_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013017.1 NP_037149.1 687 130
          GenBank mRNA DY310463.1 - 208 130
          M83676.1 - 5 - 6 482 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002629.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle docking involved in exocytosis
          autophagy
          small GTPase mediated signal transduction
          endosome to lysosome transport
          protein secretion
          protein transport
          positive regulation of macroautophagy
          regulation of exocytosis
          Rab protein signal transduction
          cellular response to insulin stimulus
          cellular protein catabolic process
          protein localization to plasma membrane
          GTPase activity
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P744999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P744999 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P744999 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P744999 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P744999 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536315 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704913 - -
          Affymetrix Rat Gene 1.0 ST Array 10930341 - -
          Affymetrix Rat Gene 1.1 ST Array 10704912 - -
          Affymetrix Rat Gene 1.1 ST Array 10930340 - -
          Rat Genome 230 2.0 Array 1371055_PM_at - -
          Rat Genome 230 2.0 Array 1371871_PM_at - -
          Rat Genome 230 2.0 Array 1371871_at - -
          Rat Genome 230 2.0 Array 1371055_at - -
          Affymetrix HT Rat Focus Array 1371055_at - -
          Affymetrix HT Rat Focus Array 1371055_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900021416.rn.1 TC0900002629.rn.1 RTA TC
          Rat Expression Set 230 1371871_at - -
          Rat Expression Set 230 1371055_PM_at - -
          Rat Expression Set 230 1371055_at - -
          Rat Expression Set 230 1371871_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704912 - -
          Affymetrix Rat Gene 1.0 ST Array 10930340 - -
          Rat Genome U34 Set M83676_at - -
          Rat Genome U34 Set rc_AI171218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21988994 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0