Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416801_m1
          See other LOC103690007 GE Assays ›
          Gene Symbol:
          LOC103690007 Nsun4
          Gene Name:
          5-methylcytosine rRNA methyltransferase NSUN4
          NOP2/Sun domain family, member 4
          Gene Aliases:
          -
          RGD1559652
          Chromosome Location:
          Chr.5: 134362953 - 134383688 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 134885377 - 134905492 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01416801_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103690007
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763980.1 XP_008762202.1 1865 96
          Gene Symbol: Nsun4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106678.1 NP_001100148.1 5 - 6 971 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004264.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          rRNA methylation
          methylation
          mature ribosome assembly
          positive regulation of mitochondrial translation
          molecular_function
          methyltransferase activity
          small ribosomal subunit rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071918 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071913 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071918 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071913 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33562 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871180 - -
          Affymetrix Rat Gene 1.0 ST Array 10871175 - -
          Affymetrix Rat Gene 1.0 ST Array 10871178 - -
          Affymetrix Rat Gene 1.0 ST Array 10871176 - -
          Affymetrix Rat Gene 1.0 ST Array 10871181 - -
          Affymetrix Rat Gene 1.0 ST Array 10871174 - -
          Affymetrix Rat Gene 1.0 ST Array 10871179 - -
          Affymetrix Rat Gene 1.1 ST Array 10871173 - -
          Rat Genome 230 2.0 Array 1373509_at - -
          Rat Genome 230 2.0 Array 1373509_PM_at - -
          Rat Genome 230 2.0 Array 1380141_at - -
          Rat Genome 230 2.0 Array 1380141_PM_at - -
          Affymetrix HT Rat Focus Array 1373509_PM_at - -
          Affymetrix HT Rat Focus Array 1380141_PM_at - -
          Affymetrix HT Rat Focus Array 1373509_at - -
          Affymetrix HT Rat Focus Array 1380141_at - -
          Affymetrix Rat Clariom S Assay PSR0500033188.rn.1 TC0500004264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500033190.rn.1 TC0500004264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500033191.rn.1 TC0500004264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500033189.rn.1 TC0500004264.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500033192.rn.1 TC0500004264.rn.1 RTA TC
          Rat Expression Set 230 1373509_at - -
          Rat Expression Set 230 1373509_PM_at - -
          Rat Expression Set 230 1380141_PM_at - -
          Rat Expression Set 230 1380141_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871173 - -
          Rat Genome U34 Set rc_AA800200_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21447426 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352671 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0