Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412821_m1
          See other GNA12 GE Assays ›
          Gene Symbol:
          Gna12
          Gene Name:
          guanine nucleotide binding protein (G protein) alpha 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 15890202 - 15971212 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412821_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gna12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031034.2 NP_112296.1 3 - 4 605 142
          GenBank mRNA D85760.1 - 3 - 4 639 142
          DQ120479.1 - 3 - 4 574 142
          DQ120480.1 - 3 - 4 574 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000270.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gna12

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          adenylate cyclase-modulating G protein-coupled receptor signaling pathway
          dopamine receptor signaling pathway
          Rho protein signal transduction
          metabolic process
          regulation of cell shape
          response to xenobiotic stimulus
          regulation of fibroblast migration
          cell differentiation
          regulation of TOR signaling
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          intracellular signal transduction
          embryonic digit morphogenesis
          GTPase activity
          GTP binding
          G-protein beta/gamma-subunit complex binding
          D5 dopamine receptor binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102467 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152963 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152963 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15312 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82960 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67764 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756902 - -
          Affymetrix Rat Gene 1.0 ST Array 10756901 - -
          Affymetrix Rat Gene 1.0 ST Array 10756904 - -
          Affymetrix Rat Gene 1.0 ST Array 10756903 - -
          Affymetrix Rat Gene 1.0 ST Array 10756900 - -
          Affymetrix Rat Gene 1.1 ST Array 10756899 - -
          Rat Genome 230 2.0 Array 1371798_at - -
          Rat Genome 230 2.0 Array 1369278_PM_at - -
          Rat Genome 230 2.0 Array 1369278_at - -
          Rat Genome 230 2.0 Array 1371798_PM_at - -
          Affymetrix HT Rat Focus Array 1369278_at - -
          Affymetrix HT Rat Focus Array 1369278_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001976.rn.1 TC1200000270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001977.rn.1 TC1200000270.rn.1 RTA TC
          Rat Expression Set 230 1369278_PM_at - -
          Rat Expression Set 230 1369278_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756899 - -
          Rat Genome U34 Set D85760_at - -
          Rat Genome U34 Set S50461_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22135944 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352707 - -
          RatRef-12 Expression BeadChip ILMN_66475 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0