Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411273_g1
          See other PDLIM1 GE Assays ›
          Gene Symbol:
          Pdlim1
          Gene Name:
          PDZ and LIM domain 1
          Gene Aliases:
          CLP36
          Chromosome Location:
          Chr.1: 259308326 - 259357006 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411273_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdlim1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017365.2 NP_059061.1 6 - 7 858 97
          GenBank mRNA BC072465.1 - 6 - 7 849 97
          FQ227333.1 - 3 - 4 377 97
          U23769.1 - 6 - 7 858 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007673.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdlim1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of transcription, DNA-templated
          transcription coactivator activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P802550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92404 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P802550 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P802550 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P802550 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P802550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5608 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23823 - -
          Agilent Rat cDNA Microarray (G4105A) 700481935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730062 - -
          Affymetrix Rat Gene 1.0 ST Array 10730059 - -
          Affymetrix Rat Gene 1.0 ST Array 10730061 - -
          Affymetrix Rat Gene 1.0 ST Array 10730060 - -
          Affymetrix Rat Gene 1.0 ST Array 10730064 - -
          Affymetrix Rat Gene 1.0 ST Array 10730063 - -
          Affymetrix Rat Gene 1.0 ST Array 10730058 - -
          Affymetrix Rat Gene 1.1 ST Array 10730057 - -
          Rat Genome 230 2.0 Array 1367946_PM_at - -
          Rat Genome 230 2.0 Array 1367946_at - -
          Affymetrix HT Rat Focus Array 1367946_PM_at - -
          Affymetrix HT Rat Focus Array 1367946_at - -
          Affymetrix Rat Clariom S Assay PSR0100061588.rn.1 TC0100007673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061584.rn.1 TC0100007673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061587.rn.1 TC0100007673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061589.rn.1 TC0100007673.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100061586.rn.1 TC0100007673.rn.1 RTA TC
          Rat Expression Set 230 1367946_PM_at - -
          Rat Expression Set 230 1367946_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730057 - -
          Rat Genome U34 Set U23769_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21345089 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361838 - -
          RatRef-12 Expression BeadChip ILMN_66007 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0