Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402090_s1
          See other GPR112L GE Assays ›
          Gene Symbol:
          Gpr112l
          Gene Name:
          G protein-coupled receptor 112 like
          Gene Aliases:
          Gpr112, RGD1564057
          Chromosome Location:
          Chr.X: 159286775 - 159437456 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402090_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr112l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773667.1 XP_008771889.1 3 6196 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059465 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059466 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059465 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059461 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155245 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166375 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P366039 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935568 - -
          Affymetrix Rat Gene 1.0 ST Array 10935569 - -
          Affymetrix Rat Gene 1.0 ST Array 10935571 - -
          Affymetrix Rat Gene 1.0 ST Array 10935574 - -
          Affymetrix Rat Gene 1.0 ST Array 10935582 - -
          Affymetrix Rat Gene 1.0 ST Array 10935580 - -
          Affymetrix Rat Gene 1.0 ST Array 10935575 - -
          Affymetrix Rat Gene 1.0 ST Array 10935572 - -
          Affymetrix Rat Gene 1.0 ST Array 10935570 - -
          Affymetrix Rat Gene 1.0 ST Array 10935581 - -
          Affymetrix Rat Gene 1.0 ST Array 10935576 - -
          Affymetrix Rat Gene 1.0 ST Array 10935573 - -
          Affymetrix Rat Gene 1.0 ST Array 10935583 - -
          Affymetrix Rat Gene 1.0 ST Array 10935577 - -
          Affymetrix Rat Gene 1.0 ST Array 10935578 - -
          Affymetrix Rat Gene 1.0 ST Array 10935566 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22329625 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351364 - -
          RatRef-12 Expression BeadChip ILMN_64053 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0