Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00690703_m1
          See other INSR GE Assays ›
          Gene Symbol:
          Insr
          Gene Name:
          insulin receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 1678620 - 1816587 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00690703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017071.2 NP_058767.2 20 - 21 4011 68
          XM_006248752.2 XP_006248814.1 20 - 21 4181 68
          XM_006248753.2 XP_006248815.1 19 - 20 4148 68
          GenBank mRNA AY566293.1 - 5 - 6 720 68
          KF112847.1 - 20 - 21 3737 68
          KF112848.1 - 20 - 21 3737 68
          M29014.1 - 20 - 21 4007 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000920.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insr

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart morphogenesis
          regulation of transcription, DNA-templated
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          G protein-coupled receptor signaling pathway
          positive regulation of cell population proliferation
          insulin receptor signaling pathway
          epidermis development
          response to hormone
          response to glucose
          animal organ morphogenesis
          response to manganese ion
          regulation of hydrogen peroxide metabolic process
          positive regulation of glycoprotein biosynthetic process
          negative regulation of gene expression
          response to activity
          peptidyl-tyrosine phosphorylation
          transformation of host cell by virus
          male sex determination
          positive regulation of cell migration
          exocrine pancreas development
          response to nutrient levels
          activation of protein kinase activity
          activation of protein kinase B activity
          response to estradiol
          negative regulation of transporter activity
          response to insulin
          cellular response to insulin stimulus
          response to vitamin D
          response to testosterone
          response to tumor necrosis factor
          peptidyl-tyrosine autophosphorylation
          positive regulation of phosphorylation
          glucose homeostasis
          positive regulation of MAPK cascade
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          regulation of embryonic development
          positive regulation of glucose import
          protein autophosphorylation
          positive regulation of developmental growth
          protein heterotetramerization
          response to glucocorticoid
          positive regulation of protein kinase B signaling
          positive regulation of respiratory burst
          cellular response to growth factor stimulus
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          insulin-activated receptor activity
          insulin-like growth factor receptor binding
          protein binding
          ATP binding
          GTP binding
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          lipoic acid binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding
          3-phosphoinositide-dependent protein kinase binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding
          protein-containing complex binding
          PTB domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118402 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71311 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24920 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756243 - -
          Affymetrix Rat Gene 1.0 ST Array 10756247 - -
          Affymetrix Rat Gene 1.0 ST Array 10756242 - -
          Affymetrix Rat Gene 1.0 ST Array 10756244 - -
          Affymetrix Rat Gene 1.0 ST Array 10756249 - -
          Affymetrix Rat Gene 1.0 ST Array 10756233 - -
          Affymetrix Rat Gene 1.0 ST Array 10756234 - -
          Affymetrix Rat Gene 1.0 ST Array 10756240 - -
          Affymetrix Rat Gene 1.0 ST Array 10756241 - -
          Affymetrix Rat Gene 1.0 ST Array 10756246 - -
          Affymetrix Rat Gene 1.0 ST Array 10756236 - -
          Affymetrix Rat Gene 1.0 ST Array 10756239 - -
          Affymetrix Rat Gene 1.0 ST Array 10756230 - -
          Affymetrix Rat Gene 1.0 ST Array 10756250 - -
          Affymetrix Rat Gene 1.0 ST Array 10756245 - -
          Affymetrix Rat Gene 1.0 ST Array 10756231 - -
          Affymetrix Rat Gene 1.0 ST Array 10756248 - -
          Affymetrix Rat Gene 1.0 ST Array 10756232 - -
          Affymetrix Rat Gene 1.0 ST Array 10756237 - -
          Affymetrix Rat Gene 1.0 ST Array 10756235 - -
          Affymetrix Rat Gene 1.0 ST Array 10756238 - -
          Affymetrix Rat Gene 1.1 ST Array 10756229 - -
          Rat Genome 230 2.0 Array 1369051_at - -
          Rat Genome 230 2.0 Array 1369051_PM_at - -
          Rat Genome 230 2.0 Array 1392043_at - -
          Rat Genome 230 2.0 Array 1392043_PM_at - -
          Affymetrix HT Rat Focus Array 1369051_at - -
          Affymetrix HT Rat Focus Array 1369051_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200007994.rn.1 TC1200000920.rn.1 RTA TC
          Rat Expression Set 230 1369051_PM_at - -
          Rat Expression Set 230 1369051_at - -
          Rat Expression Set 230 1392043_at - -
          Rat Expression Set 230 1392043_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756229 - -
          Rat Genome U34 Set M29014_at - -
          Rat Genome U34 Set M29014_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22333797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360127 - -
          RatRef-12 Expression BeadChip ILMN_67126 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header