Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00690189_g1
          See other AVP GE Assays ›
          Gene Symbol:
          Avp
          Gene Name:
          arginine vasopressin
          Gene Aliases:
          DI, VP, Vas
          Chromosome Location:
          Chr.3: 123117482 - 123119460 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00690189_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016992.2 NP_058688.2 2 - 3 357 78
          GenBank mRNA M25646.1 - 2 - 3 357 78
          M64785.1 - 705 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004172.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Avp

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          maternal aggressive behavior
          positive regulation of systemic arterial blood pressure
          protein phosphorylation
          smooth muscle contraction
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          negative regulation of female receptivity
          grooming behavior
          locomotory behavior
          positive regulation of cell population proliferation
          positive regulation of gene expression
          positive regulation of glutamate secretion
          response to organic cyclic compound
          positive regulation of cell growth
          positive regulation of prostaglandin biosynthetic process
          positive regulation of cellular pH reduction
          positive regulation of peptidyl-serine phosphorylation
          response to testosterone
          response to nicotine
          social behavior
          regulation of renal sodium excretion
          vasoconstriction
          hyperosmotic salinity response
          maternal behavior
          negative regulation of apoptotic process
          penile erection
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to ethanol
          positive regulation of vasoconstriction
          multicellular organismal water homeostasis
          negative regulation of transmission of nerve impulse
          ERK1 and ERK2 cascade
          protein kinase C signaling
          negative regulation of release of cytochrome c from mitochondria
          protein kinase activity
          neuropeptide hormone activity
          neurohypophyseal hormone activity
          protein binding
          V1A vasopressin receptor binding
          V1B vasopressin receptor binding
          V2 vasopressin receptor binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P254016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P254016 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P254016 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P254016 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P254016 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452171 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700489075 - -
          Agilent Rat cDNA Microarray (G4105A) 700489075 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10849917 - -
          Affymetrix Rat Gene 1.0 ST Array 10849916 - -
          Affymetrix Rat Gene 1.1 ST Array 10849915 - -
          Rat Genome 230 2.0 Array 1387317_PM_at - -
          Rat Genome 230 2.0 Array 1387317_at - -
          Affymetrix HT Rat Focus Array 1387317_at - -
          Affymetrix HT Rat Focus Array 1387317_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300032393.rn.1 TC0300004172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300032394.rn.1 TC0300004172.rn.1 RTA TC
          Rat Expression Set 230 1387317_PM_at - -
          Rat Expression Set 230 1387317_at - -
          Affymetrix Rat Gene 1.0 ST Array 10849915 - -
          Rat Genome U34 Set M64785_g_at - -
          Rat Genome U34 Set M64785_at - -
          Rat Genome U34 Set M25646_at - -
          Rat Genome U34 Set M64785_g_at - -
          Rat Genome U34 Set M25646_at - -
          Rat Genome U34 Set M64785_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21004736 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372854 - -
          RatRef-12 Expression BeadChip ILMN_53350 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0