Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00672126_m1
          See other JDP2 GE Assays ›
          Gene Symbol:
          Jdp2
          Gene Name:
          Jun dimerization protein 2
          Gene Aliases:
          Jundm2, Jundp2
          Chromosome Location:
          Chr.6: 109464908 - 109505440 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00672126_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jdp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053894.1 NP_446346.1 2 - 3 576 65
          XM_008764734.1 XP_008762956.1 3 - 4 1146 65
          XM_008764735.1 XP_008762957.1 3 - 4 473 65
          GenBank mRNA U53449.1 - 2 - 3 576 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003484.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jdp2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of histone deacetylation
          negative regulation of fat cell differentiation
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P508466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P508466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886038 - -
          Affymetrix Rat Gene 1.0 ST Array 10886039 - -
          Affymetrix Rat Gene 1.0 ST Array 10886037 - -
          Affymetrix Rat Gene 1.1 ST Array 10886036 - -
          Rat Genome 230 2.0 Array 1372986_at - -
          Rat Genome 230 2.0 Array 1369583_PM_at - -
          Rat Genome 230 2.0 Array 1369583_at - -
          Rat Genome 230 2.0 Array 1372986_PM_at - -
          Affymetrix HT Rat Focus Array 1372986_PM_at - -
          Affymetrix HT Rat Focus Array 1369583_at - -
          Affymetrix HT Rat Focus Array 1369583_PM_at - -
          Affymetrix HT Rat Focus Array 1372986_at - -
          Affymetrix Rat Clariom S Assay PSR0600024459.rn.1 TC0600003484.rn.1 RTA TC
          Rat Expression Set 230 1372986_at - -
          Rat Expression Set 230 1369583_at - -
          Rat Expression Set 230 1369583_PM_at - -
          Rat Expression Set 230 1372986_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886036 - -
          Rat Genome U34 Set U53449_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21192056 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371286 - -
          RatRef-12 Expression BeadChip ILMN_62201 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0