Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591638_m1
          See other ZFP384 GE Assays ›
          Gene Symbol:
          Zfp384
          Gene Name:
          zinc finger protein 384
          Gene Aliases:
          Ciz, Nmp4, Znf384
          Chromosome Location:
          Chr.4: 157523320 - 157552606 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591638_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp384
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034830.1 NP_001030002.1 6 - 7 891 102
          NM_001034831.1 NP_001030003.1 6 - 7 890 102
          NM_133429.2 NP_596920.2 6 - 7 974 102
          XM_006237328.2 XP_006237390.1 4 - 5 687 102
          GenBank mRNA AB019281.1 - 6 - 7 891 102
          AF216804.1 - 6 - 7 974 102
          AF216805.1 - 4 - 5 693 102
          AF216806.1 - 6 - 7 890 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001781.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp384

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          nucleocytoplasmic transport
          positive regulation of transcription by RNA polymerase II
          positive regulation of protein secretion
          nucleic acid binding
          SH3 domain binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P374902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156484 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P374902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491653 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70883 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858750 - -
          Affymetrix Rat Gene 1.0 ST Array 10858739 - -
          Affymetrix Rat Gene 1.0 ST Array 10858737 - -
          Affymetrix Rat Gene 1.0 ST Array 10858738 - -
          Affymetrix Rat Gene 1.0 ST Array 10858742 - -
          Affymetrix Rat Gene 1.0 ST Array 10858736 - -
          Affymetrix Rat Gene 1.0 ST Array 10858743 - -
          Affymetrix Rat Gene 1.0 ST Array 10858744 - -
          Affymetrix Rat Gene 1.0 ST Array 10858749 - -
          Affymetrix Rat Gene 1.0 ST Array 10858740 - -
          Affymetrix Rat Gene 1.0 ST Array 10858748 - -
          Affymetrix Rat Gene 1.0 ST Array 10858741 - -
          Affymetrix Rat Gene 1.0 ST Array 10858746 - -
          Affymetrix Rat Gene 1.0 ST Array 10858751 - -
          Affymetrix Rat Gene 1.0 ST Array 10858747 - -
          Affymetrix Rat Gene 1.1 ST Array 10858734 - -
          Rat Genome 230 2.0 Array 1370175_a_at - -
          Rat Genome 230 2.0 Array 1370175_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370175_a_at - -
          Affymetrix HT Rat Focus Array 1370175_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400012901.rn.1 TC0400001781.rn.1 RTA TC
          Rat Expression Set 230 1370175_a_at - -
          Rat Expression Set 230 1370175_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858734 - -
          Rat Genome U34 Set rc_AI177799_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21411205 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356146 - -
          RatRef-12 Expression BeadChip ILMN_68598 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0