Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590820_m1
          See other RASGRP4 GE Assays ›
          Gene Symbol:
          Rasgrp4
          Gene Name:
          RAS guanyl releasing protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 88078350 - 88094331 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590820_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasgrp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130824.2 NP_570837.2 5 - 6 666 68
          GenBank mRNA AF465263.1 - 5 - 6 666 68
          AF465264.1 - 5 - 6 666 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001215.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasgrp4

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          cell population proliferation
          response to extracellular stimulus
          cell growth
          cell differentiation
          positive regulation of Ras protein signal transduction
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32587 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007074 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007074 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007074 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705727 - -
          Affymetrix Rat Gene 1.0 ST Array 10705717 - -
          Affymetrix Rat Gene 1.0 ST Array 10705713 - -
          Affymetrix Rat Gene 1.0 ST Array 10705718 - -
          Affymetrix Rat Gene 1.0 ST Array 10705719 - -
          Affymetrix Rat Gene 1.0 ST Array 10705714 - -
          Affymetrix Rat Gene 1.0 ST Array 10705715 - -
          Affymetrix Rat Gene 1.0 ST Array 10705720 - -
          Affymetrix Rat Gene 1.0 ST Array 10705728 - -
          Affymetrix Rat Gene 1.0 ST Array 10705721 - -
          Affymetrix Rat Gene 1.0 ST Array 10705724 - -
          Affymetrix Rat Gene 1.0 ST Array 10705725 - -
          Affymetrix Rat Gene 1.0 ST Array 10705726 - -
          Affymetrix Rat Gene 1.0 ST Array 10705723 - -
          Affymetrix Rat Gene 1.0 ST Array 10705716 - -
          Affymetrix Rat Gene 1.0 ST Array 10705722 - -
          Affymetrix Rat Gene 1.1 ST Array 10705712 - -
          Rat Genome 230 2.0 Array 1368781_at - -
          Rat Genome 230 2.0 Array 1368781_PM_at - -
          Affymetrix HT Rat Focus Array 1368781_at - -
          Affymetrix HT Rat Focus Array 1368781_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100009349.rn.1 TC0100001215.rn.1 RTA TC
          Rat Expression Set 230 1368781_at - -
          Rat Expression Set 230 1368781_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705712 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21766212 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650789 - -
          RatRef-12 Expression BeadChip ILMN_62191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-v5hlb:80/100.66.134.185:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0