Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589991_m1
          See other COPB1 GE Assays ›
          Gene Symbol:
          Copb1
          Gene Name:
          coatomer protein complex, subunit beta 1
          Gene Aliases:
          Rack2
          Chromosome Location:
          Chr.1: 183695480 - 183729569 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589991_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Copb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080781.2 NP_542959.1 4 - 5 645 67
          XM_008759660.1 XP_008757882.1 4 - 5 648 67
          XM_008759661.1 XP_008757883.1 4 - 5 735 67
          GenBank mRNA BC061882.1 - 4 - 5 645 67
          X57228.1 - 4 - 5 576 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008128.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Copb1

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
          intra-Golgi vesicle-mediated transport
          structural molecule activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P455785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42583 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P455785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P455785 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P455785 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P455785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725005 - -
          Affymetrix Rat Gene 1.0 ST Array 10725013 - -
          Affymetrix Rat Gene 1.0 ST Array 10725012 - -
          Affymetrix Rat Gene 1.0 ST Array 10725020 - -
          Affymetrix Rat Gene 1.0 ST Array 10725014 - -
          Affymetrix Rat Gene 1.0 ST Array 10725015 - -
          Affymetrix Rat Gene 1.0 ST Array 10725024 - -
          Affymetrix Rat Gene 1.0 ST Array 10725009 - -
          Affymetrix Rat Gene 1.0 ST Array 10725010 - -
          Affymetrix Rat Gene 1.0 ST Array 10725008 - -
          Affymetrix Rat Gene 1.0 ST Array 10725017 - -
          Affymetrix Rat Gene 1.0 ST Array 10725018 - -
          Affymetrix Rat Gene 1.0 ST Array 10725025 - -
          Affymetrix Rat Gene 1.0 ST Array 10725023 - -
          Affymetrix Rat Gene 1.0 ST Array 10725016 - -
          Affymetrix Rat Gene 1.0 ST Array 10725011 - -
          Affymetrix Rat Gene 1.0 ST Array 10725026 - -
          Affymetrix Rat Gene 1.0 ST Array 10725021 - -
          Affymetrix Rat Gene 1.0 ST Array 10725006 - -
          Affymetrix Rat Gene 1.0 ST Array 10725007 - -
          Affymetrix Rat Gene 1.0 ST Array 10725019 - -
          Affymetrix Rat Gene 1.0 ST Array 10725022 - -
          Affymetrix Rat Gene 1.1 ST Array 10725004 - -
          Rat Genome 230 2.0 Array 1367461_PM_at - -
          Rat Genome 230 2.0 Array 1367461_at - -
          Affymetrix HT Rat Focus Array 1367461_PM_at - -
          Affymetrix HT Rat Focus Array 1367461_at - -
          Affymetrix Rat Clariom S Assay PSR0100066813.rn.1 TC0100008128.rn.1 RTA TC
          Rat Expression Set 230 1367461_at - -
          Rat Expression Set 230 1367461_PM_at - -
          Rat Expression Set 230 1367461_PM_at - -
          Rat Expression Set 230 1367461_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725004 - -
          Rat Genome U34 Set rc_AI105054_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20729056 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363677 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039866 - -
          RatRef-12 Expression BeadChip ILMN_161711 - -
          RatRef-12 Expression BeadChip ILMN_68176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0