Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582384_m1
          See other ATP6AP1 GE Assays ›
          Gene Symbol:
          Atp6ap1
          Gene Name:
          ATPase, H+ transporting, lysosomal accessory protein 1
          Gene Aliases:
          Atp6s1, C7-1
          Chromosome Location:
          Chr.X: 156407973 - 156415053 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6ap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031785.1 NP_113973.1 7 - 8 903 61
          GenBank mRNA AF035387.1 - 7 - 8 903 61
          BC070937.1 - 7 - 8 903 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002633.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6ap1

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          cell death
          positive regulation of osteoblast differentiation
          positive regulation of bone resorption
          pH reduction
          positive regulation of exocytosis
          establishment of organelle localization
          proton-transporting V-type ATPase complex assembly
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of osteoclast development
          molecular_function
          ATP binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105076 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12679 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12679 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12679 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700519286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7459 - -
          Agilent Rat cDNA Microarray (G4105A) 700519286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940387 - -
          Affymetrix Rat Gene 1.0 ST Array 10940385 - -
          Affymetrix Rat Gene 1.0 ST Array 10940388 - -
          Affymetrix Rat Gene 1.0 ST Array 10940389 - -
          Affymetrix Rat Gene 1.0 ST Array 10940386 - -
          Affymetrix Rat Gene 1.0 ST Array 10940390 - -
          Affymetrix Rat Gene 1.0 ST Array 10940382 - -
          Affymetrix Rat Gene 1.0 ST Array 10940381 - -
          Affymetrix Rat Gene 1.0 ST Array 10940383 - -
          Affymetrix Rat Gene 1.0 ST Array 10940384 - -
          Affymetrix Rat Gene 1.1 ST Array 10940380 - -
          Rat Genome 230 2.0 Array 1367873_at - -
          Rat Genome 230 2.0 Array 1367873_PM_at - -
          Affymetrix HT Rat Focus Array 1367873_at - -
          Affymetrix HT Rat Focus Array 1367873_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016334.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016339.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016331.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016340.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016344.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016329.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016335.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016337.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016345.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016330.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016341.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016332.rn.1 TC0X00002633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016333.rn.1 TC0X00002633.rn.1 RTA TC
          Rat Expression Set 230 1367873_at - -
          Rat Expression Set 230 1367873_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940380 - -
          Rat Genome U34 Set AF035387_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21307314 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359644 - -
          RatRef-12 Expression BeadChip ILMN_56364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0