Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581426_m1
          See other CAMKK1 GE Assays ›
          Gene Symbol:
          Camkk1
          Gene Name:
          calcium/calmodulin-dependent protein kinase kinase 1, alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 59585034 - 59608180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581426_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camkk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031662.1 NP_113850.1 9 - 10 903 80
          XM_006246799.2 XP_006246861.1 10 - 11 1103 80
          XM_006246801.2 XP_006246863.1 10 - 11 1050 80
          XM_008767866.1 XP_008766088.1 11 - 12 1192 80
          GenBank mRNA AB023658.1 - 9 - 10 903 80
          L42810.1 - 9 - 10 903 80
          S83194.1 - 9 - 10 903 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001019.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camkk1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          activation of protein kinase activity
          positive regulation of protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calmodulin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P322860 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171136 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P322860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P322860 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P322860 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103790 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P322860 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700325250 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701714505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63967 - -
          Agilent Rat cDNA Microarray (G4105A) 700325250 - -
          Agilent Rat cDNA Microarray (G4105A) 701714505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735623 - -
          Affymetrix Rat Gene 1.0 ST Array 10735632 - -
          Affymetrix Rat Gene 1.0 ST Array 10735628 - -
          Affymetrix Rat Gene 1.0 ST Array 10735630 - -
          Affymetrix Rat Gene 1.0 ST Array 10735620 - -
          Affymetrix Rat Gene 1.0 ST Array 10735626 - -
          Affymetrix Rat Gene 1.0 ST Array 10735622 - -
          Affymetrix Rat Gene 1.0 ST Array 10735625 - -
          Affymetrix Rat Gene 1.0 ST Array 10735627 - -
          Affymetrix Rat Gene 1.0 ST Array 10735616 - -
          Affymetrix Rat Gene 1.0 ST Array 10735631 - -
          Affymetrix Rat Gene 1.0 ST Array 10735619 - -
          Affymetrix Rat Gene 1.0 ST Array 10735629 - -
          Affymetrix Rat Gene 1.0 ST Array 10735618 - -
          Affymetrix Rat Gene 1.0 ST Array 10735621 - -
          Affymetrix Rat Gene 1.0 ST Array 10735617 - -
          Affymetrix Rat Gene 1.1 ST Array 10735615 - -
          Rat Genome 230 2.0 Array 1368156_PM_at - -
          Rat Genome 230 2.0 Array 1368156_at - -
          Affymetrix HT Rat Focus Array 1368156_at - -
          Affymetrix HT Rat Focus Array 1368156_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000009294.rn.1 TC1000001019.rn.1 RTA TC
          Rat Expression Set 230 1368156_PM_at - -
          Rat Expression Set 230 1368156_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735615 - -
          Rat Genome U34 Set S83194_s_at - -
          Rat Genome U34 Set S83194_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21698502 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351458 - -
          RatRef-12 Expression BeadChip ILMN_56724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0