Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575742_m1
          See other CSNK1G3 GE Assays ›
          Gene Symbol:
          Csnk1g3
          Gene Name:
          casein kinase 1, gamma 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 48831209 - 48913341 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575742_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csnk1g3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022855.2 NP_074046.1 9 - 10 1846 108

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csnk1g3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          endocytosis
          regulation of cell shape
          Wnt signaling pathway
          peptidyl-serine phosphorylation
          protein autophosphorylation
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          peptide binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P105004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105004 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105004 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P105004 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P105004 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17678 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881460 - -
          Affymetrix Rat Gene 1.0 ST Array 10881459 - -
          Affymetrix Rat Gene 1.0 ST Array 10801772 - -
          Affymetrix Rat Gene 1.0 ST Array 10801774 - -
          Affymetrix Rat Gene 1.0 ST Array 10881467 - -
          Affymetrix Rat Gene 1.0 ST Array 10801777 - -
          Affymetrix Rat Gene 1.0 ST Array 10801776 - -
          Affymetrix Rat Gene 1.0 ST Array 10801770 - -
          Affymetrix Rat Gene 1.0 ST Array 10801775 - -
          Affymetrix Rat Gene 1.0 ST Array 10801773 - -
          Affymetrix Rat Gene 1.0 ST Array 10801771 - -
          Affymetrix Rat Gene 1.1 ST Array 10881465 - -
          Affymetrix Rat Gene 1.1 ST Array 10801769 - -
          Affymetrix Rat Gene 1.1 ST Array 10881456 - -
          Rat Genome 230 2.0 Array 1382574_PM_at - -
          Rat Genome 230 2.0 Array 1368849_PM_at - -
          Rat Genome 230 2.0 Array 1368849_at - -
          Rat Genome 230 2.0 Array 1368850_at - -
          Rat Genome 230 2.0 Array 1394731_at - -
          Rat Genome 230 2.0 Array 1372582_at - -
          Rat Genome 230 2.0 Array 1372582_PM_at - -
          Rat Genome 230 2.0 Array 1394731_PM_at - -
          Rat Genome 230 2.0 Array 1368850_PM_at - -
          Rat Genome 230 2.0 Array 1382574_at - -
          Affymetrix HT Rat Focus Array 1394731_PM_at - -
          Affymetrix HT Rat Focus Array 1372582_PM_at - -
          Affymetrix HT Rat Focus Array 1368849_at - -
          Affymetrix HT Rat Focus Array 1394731_at - -
          Affymetrix HT Rat Focus Array 1368850_at - -
          Affymetrix HT Rat Focus Array 1368849_PM_at - -
          Affymetrix HT Rat Focus Array 1372582_at - -
          Affymetrix HT Rat Focus Array 1368850_PM_at - -
          Rat Expression Set 230 1368850_PM_at - -
          Rat Expression Set 230 1372582_PM_at - -
          Rat Expression Set 230 1368850_at - -
          Rat Expression Set 230 1368849_at - -
          Rat Expression Set 230 1372582_at - -
          Rat Expression Set 230 1368849_PM_at - -
          Rat Expression Set 230 1394731_at - -
          Rat Expression Set 230 1382574_at - -
          Rat Expression Set 230 1394731_PM_at - -
          Rat Expression Set 230 1382574_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801769 - -
          Rat Genome U34 Set U22321_at - -
          Rat Genome U34 Set rc_AI176776_at - -
          Rat Genome U34 Set rc_AI178699_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20715317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365125 - -
          RatRef-12 Expression BeadChip ILMN_63083 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0