Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574684_m1
          See other CASP2 GE Assays ›
          Gene Symbol:
          Casp2
          Gene Name:
          caspase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 71652362 - 71670228 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574684_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022522.2 NP_071967.2 6 - 7 749 86
          XM_006236394.2 XP_006236456.1 5 - 6 1430 86
          XM_008762918.1 XP_008761140.1 5 - 6 1430 86
          GenBank mRNA AF025671.1 - 5 - 6 542 86
          AF136231.1 - 6 - 7 747 86
          AF136232.1 - 6 - 7 747 86
          U34684.1 - 1 - 2 152 86
          U77933.1 - 6 - 7 749 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000752.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp2

          Gene Ontology Categories:

          Function(s) Process(es)

          luteolysis
          neural retina development
          proteolysis
          apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          brain development
          aging
          intrinsic apoptotic signaling pathway in response to DNA damage
          programmed cell death
          protein processing
          ectopic germ cell programmed cell death
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of neuron apoptotic process
          cellular response to mechanical stimulus
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of apoptotic signaling pathway
          molecular_function
          cysteine-type endopeptidase activity
          protein binding
          protein domain specific binding
          cysteine-type endopeptidase activity involved in apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164041 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158644 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1019923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019923 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164041 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019923 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164041 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1019923 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164041 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158644 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019923 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700144907 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30096 - -
          Agilent Rat cDNA Microarray (G4105A) 700144907 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855027 - -
          Affymetrix Rat Gene 1.0 ST Array 10855026 - -
          Affymetrix Rat Gene 1.0 ST Array 10855032 - -
          Affymetrix Rat Gene 1.0 ST Array 10855024 - -
          Affymetrix Rat Gene 1.0 ST Array 10855031 - -
          Affymetrix Rat Gene 1.0 ST Array 10855021 - -
          Affymetrix Rat Gene 1.0 ST Array 10855034 - -
          Affymetrix Rat Gene 1.0 ST Array 10855022 - -
          Affymetrix Rat Gene 1.0 ST Array 10855025 - -
          Affymetrix Rat Gene 1.0 ST Array 10855023 - -
          Affymetrix Rat Gene 1.0 ST Array 10855030 - -
          Affymetrix Rat Gene 1.0 ST Array 10855033 - -
          Affymetrix Rat Gene 1.0 ST Array 10855028 - -
          Affymetrix Rat Gene 1.1 ST Array 10855020 - -
          Rat Genome 230 2.0 Array 1367891_PM_a_at - -
          Rat Genome 230 2.0 Array 1367890_PM_at - -
          Rat Genome 230 2.0 Array 1367890_at - -
          Rat Genome 230 2.0 Array 1367891_a_at - -
          Affymetrix HT Rat Focus Array 1367890_PM_at - -
          Affymetrix HT Rat Focus Array 1367890_at - -
          Affymetrix HT Rat Focus Array 1367891_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367891_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400005535.rn.1 TC0400000752.rn.1 RTA TC
          Rat Expression Set 230 1367891_a_at - -
          Rat Expression Set 230 1367890_PM_at - -
          Rat Expression Set 230 1367891_PM_a_at - -
          Rat Expression Set 230 1367890_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855020 - -
          Rat Genome U34 Set U34684_s_at - -
          Rat Genome U34 Set AF025671_s_at - -
          Rat Genome U34 Set U77933_at - -
          Rat Genome U34 Set AF025671_s_at - -
          Rat Genome U34 Set U34684_s_at - -
          Rat Genome U34 Set U77933_at - -
          Rat Genome U34 Set U34684_s_at - -
          Rat Genome U34 Set U77933_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22061218 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349906 - -
          RatRef-12 Expression BeadChip ILMN_66477 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0