Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573997_m1
          See other FBXL20 GE Assays ›
          Gene Symbol:
          Fbxl20
          Gene Name:
          F-box and leucine-rich repeat protein 20
          Gene Aliases:
          Fbl2, Fbxl2
          Chromosome Location:
          Chr.10: 86036745 - 86099696 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573997_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxl20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022272.1 NP_071608.1 7 - 8 493 89
          XM_006247421.2 XP_006247483.1 7 - 8 795 89
          XM_008768110.1 XP_008766332.1 7 - 8 797 89
          XM_008768111.1 XP_008766333.1 7 - 8 832 89
          GenBank mRNA AF182443.1 - 7 - 8 493 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003704.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxl20

          Gene Ontology Categories:

          Function(s)

          behavioral fear response

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P357089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P357089 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P357089 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P357089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P357089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29798 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746870 - -
          Affymetrix Rat Gene 1.0 ST Array 10746863 - -
          Affymetrix Rat Gene 1.0 ST Array 10746868 - -
          Affymetrix Rat Gene 1.0 ST Array 10746862 - -
          Affymetrix Rat Gene 1.0 ST Array 10746869 - -
          Affymetrix Rat Gene 1.0 ST Array 10746864 - -
          Affymetrix Rat Gene 1.0 ST Array 10746866 - -
          Affymetrix Rat Gene 1.0 ST Array 10746865 - -
          Affymetrix Rat Gene 1.0 ST Array 10746871 - -
          Affymetrix Rat Gene 1.0 ST Array 10746867 - -
          Affymetrix Rat Gene 1.1 ST Array 10746856 - -
          Rat Genome 230 2.0 Array 1396845_PM_at - -
          Rat Genome 230 2.0 Array 1370099_at - -
          Rat Genome 230 2.0 Array 1396845_at - -
          Rat Genome 230 2.0 Array 1370099_PM_at - -
          Affymetrix HT Rat Focus Array 1396845_PM_at - -
          Affymetrix HT Rat Focus Array 1396845_at - -
          Affymetrix HT Rat Focus Array 1370099_PM_at - -
          Affymetrix HT Rat Focus Array 1370099_at - -
          Affymetrix Rat Clariom S Assay PSR1000033123.rn.1 TC1000003704.rn.1 RTA TC
          Rat Expression Set 230 1370099_PM_at - -
          Rat Expression Set 230 1370099_at - -
          Rat Expression Set 230 1396845_at - -
          Rat Expression Set 230 1396845_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746856 - -
          Rat Genome U34 Set rc_AI170605_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21946206 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355110 - -
          RatRef-12 Expression BeadChip ILMN_57724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0