Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570328_m1
          See other MGAT3 GE Assays ›
          Gene Symbol:
          Mgat3
          Gene Name:
          mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121394541 - 121436935 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570328_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgat3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019239.2 NP_062112.2 1 - 2 50 96
          GenBank mRNA D10852.1 - 1 - 2 50 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001474.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the D10852.1, NM_019239.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mgat3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          beta-1,4-mannosylglycoprotein 4-beta-N-acetylglucosaminyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160848 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072189 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33147 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89048 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897754 - -
          Affymetrix Rat Gene 1.0 ST Array 10897753 - -
          Affymetrix Rat Gene 1.1 ST Array 10897752 - -
          Rat Genome 230 2.0 Array 1369201_PM_at - -
          Rat Genome 230 2.0 Array 1369201_at - -
          Affymetrix HT Rat Focus Array 1369201_at - -
          Affymetrix HT Rat Focus Array 1369201_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700011042.rn.1 TC0700001474.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011046.rn.1 TC0700001474.rn.1 RTA TC
          Rat Expression Set 230 1369201_PM_at - -
          Rat Expression Set 230 1369201_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897752 - -
          Rat Genome U34 Set D10852_at - -
          Rat Genome U34 Set D10852_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20775257 - -
          ABI Rat Genome Survey Microarray 22290191 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351382 - -
          RatRef-12 Expression BeadChip ILMN_52246 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0