Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568783_m1
          See other GABRR2 GE Assays ›
          Gene Symbol:
          Gabrr2
          Gene Name:
          gamma-aminobutyric acid (GABA) A receptor, rho 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 48298100 - 48341695 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568783_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabrr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017292.2 NP_058988.1 8 - 9 1250 86
          XM_008763610.1 XP_008761832.1 10 - 11 1236 86
          XM_008763611.1 XP_008761833.1 7 - 8 2182 86
          GenBank mRNA D38494.1 - 8 - 9 1250 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000403.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gabrr2

          Gene Ontology Categories:

          Function(s) Process(es)

          gamma-aminobutyric acid signaling pathway
          visual perception
          ion transmembrane transport
          eye photoreceptor cell development
          chloride transmembrane transport
          GABA-A receptor activity
          inhibitory extracellular ligand-gated ion channel activity
          chloride channel activity
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217465 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15024 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868143 - -
          Affymetrix Rat Gene 1.0 ST Array 10868145 - -
          Affymetrix Rat Gene 1.0 ST Array 10868150 - -
          Affymetrix Rat Gene 1.0 ST Array 10868144 - -
          Affymetrix Rat Gene 1.0 ST Array 10868146 - -
          Affymetrix Rat Gene 1.0 ST Array 10868148 - -
          Affymetrix Rat Gene 1.0 ST Array 10868149 - -
          Affymetrix Rat Gene 1.0 ST Array 10868147 - -
          Affymetrix Rat Gene 1.1 ST Array 10868142 - -
          Rat Genome 230 2.0 Array 1369231_at - -
          Rat Genome 230 2.0 Array 1369230_at - -
          Rat Genome 230 2.0 Array 1369231_PM_at - -
          Rat Genome 230 2.0 Array 1369230_PM_at - -
          Affymetrix HT Rat Focus Array 1369231_PM_at - -
          Affymetrix HT Rat Focus Array 1369231_at - -
          Affymetrix HT Rat Focus Array 1369230_PM_at - -
          Affymetrix HT Rat Focus Array 1369230_at - -
          Affymetrix Rat Clariom S Assay PSR0500002506.rn.1 TC0500000403.rn.1 RTA TC
          Rat Expression Set 230 1369231_at - -
          Rat Expression Set 230 1369230_PM_at - -
          Rat Expression Set 230 1369230_at - -
          Rat Expression Set 230 1369231_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868142 - -
          Rat Genome U34 Set D38494_at - -
          Rat Genome U34 Set D38494_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21739591 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366995 - -
          RatRef-12 Expression BeadChip ILMN_61993 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-69f987f899-9tng4:80/100.66.130.250:80.
          git-commit: eb1b8c2b8eec225754209f035b16fef014668c1b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0