Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567413_g1
          See other GRN GE Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 90377103 - 90383207 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567413_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145842.1 NP_001139314.1 7 - 8 764 56
          NM_017113.2 NP_058809.2 7 - 8 764 56
          XM_008767981.1 XP_008766203.1 7 - 8 845 56
          GenBank mRNA BC072469.1 - 7 - 8 758 56
          M97750.1 - 6 - 7 719 56
          X62322.1 - 7 - 8 738 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001688.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grn

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          RNA binding
          cytokine activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047527 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1028858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67550 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1028858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1028858 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1028858 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047527 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1028858 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700480457 - -
          Agilent Rat cDNA Microarray (G4105A) 700480457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738585 - -
          Affymetrix Rat Gene 1.0 ST Array 10738589 - -
          Affymetrix Rat Gene 1.0 ST Array 10738584 - -
          Affymetrix Rat Gene 1.0 ST Array 10738590 - -
          Affymetrix Rat Gene 1.0 ST Array 10738578 - -
          Affymetrix Rat Gene 1.0 ST Array 10738583 - -
          Affymetrix Rat Gene 1.0 ST Array 10738582 - -
          Affymetrix Rat Gene 1.0 ST Array 10738577 - -
          Affymetrix Rat Gene 1.0 ST Array 10738586 - -
          Affymetrix Rat Gene 1.0 ST Array 10738581 - -
          Affymetrix Rat Gene 1.0 ST Array 10738580 - -
          Affymetrix Rat Gene 1.0 ST Array 10738588 - -
          Affymetrix Rat Gene 1.0 ST Array 10738587 - -
          Affymetrix Rat Gene 1.0 ST Array 10738579 - -
          Affymetrix Rat Gene 1.1 ST Array 10738576 - -
          Rat Genome 230 2.0 Array 1386893_PM_at - -
          Rat Genome 230 2.0 Array 1386893_at - -
          Affymetrix HT Rat Focus Array 1386893_at - -
          Affymetrix HT Rat Focus Array 1386893_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000015320.rn.1 TC1000001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015322.rn.1 TC1000001688.rn.1 RTA TC
          Rat Expression Set 230 1386893_PM_at - -
          Rat Expression Set 230 1386893_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738576 - -
          Rat Genome U34 Set X62322_at - -
          Rat Genome U34 Set X62322_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21842753 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357411 - -
          RatRef-12 Expression BeadChip ILMN_56067 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0