Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563181_m1
          See other ANXA3 GE Assays ›
          Gene Symbol:
          Anxa3
          Gene Name:
          annexin A3
          Gene Aliases:
          Anx3, LC3, LRRGT00047
          Chromosome Location:
          Chr.14: 14371921 - 14426507 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563181_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anxa3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012823.2 NP_036955.2 10 - 11 867 116
          XM_006250694.2 XP_006250756.1 10 - 11 1231 116
          XM_006250696.2 XP_006250758.1 11 - 12 987 116
          GenBank mRNA BC081856.1 - 9 - 10 779 116
          M20559.1 - 10 - 11 815 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001444.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anxa3

          Gene Ontology Categories:

          Function(s) Process(es)

          phagocytosis
          positive regulation of endothelial cell migration
          hippocampus development
          animal organ regeneration
          defense response to bacterium
          negative regulation of catalytic activity
          neutrophil degranulation
          positive regulation of angiogenesis
          positive regulation of DNA metabolic process
          positive regulation of DNA-binding transcription factor activity
          response to glucocorticoid
          response to growth factor
          phospholipase inhibitor activity
          calcium ion binding
          calcium-dependent phospholipid binding
          phospholipase A2 inhibitor activity
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P325508 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116690 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116690 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325508 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P325508 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116690 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P301936 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P325508 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775633 - -
          Affymetrix Rat Gene 1.0 ST Array 10775638 - -
          Affymetrix Rat Gene 1.0 ST Array 10775642 - -
          Affymetrix Rat Gene 1.0 ST Array 10775641 - -
          Affymetrix Rat Gene 1.0 ST Array 10775644 - -
          Affymetrix Rat Gene 1.0 ST Array 10775631 - -
          Affymetrix Rat Gene 1.0 ST Array 10775643 - -
          Affymetrix Rat Gene 1.0 ST Array 10775634 - -
          Affymetrix Rat Gene 1.0 ST Array 10775645 - -
          Affymetrix Rat Gene 1.0 ST Array 10775640 - -
          Affymetrix Rat Gene 1.0 ST Array 10775636 - -
          Affymetrix Rat Gene 1.0 ST Array 10775639 - -
          Affymetrix Rat Gene 1.1 ST Array 10775628 - -
          Rat Genome 230 2.0 Array 1380751_at - -
          Rat Genome 230 2.0 Array 1395313_s_at - -
          Rat Genome 230 2.0 Array 1367974_at - -
          Rat Genome 230 2.0 Array 1395313_PM_s_at - -
          Rat Genome 230 2.0 Array 1367974_PM_at - -
          Rat Genome 230 2.0 Array 1367975_at - -
          Rat Genome 230 2.0 Array 1380751_PM_at - -
          Rat Genome 230 2.0 Array 1367975_PM_at - -
          Affymetrix HT Rat Focus Array 1395313_s_at - -
          Affymetrix HT Rat Focus Array 1367974_at - -
          Affymetrix HT Rat Focus Array 1395313_PM_s_at - -
          Affymetrix HT Rat Focus Array 1367975_PM_at - -
          Affymetrix HT Rat Focus Array 1367975_at - -
          Affymetrix HT Rat Focus Array 1367974_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400010555.rn.1 TC1400001444.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010552.rn.1 TC1400001444.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010551.rn.1 TC1400001444.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400010557.rn.1 TC1400001444.rn.1 RTA TC
          Rat Expression Set 230 1380751_at - -
          Rat Expression Set 230 1367975_PM_at - -
          Rat Expression Set 230 1380751_PM_at - -
          Rat Expression Set 230 1367975_at - -
          Rat Expression Set 230 1367974_at - -
          Rat Expression Set 230 1367974_PM_at - -
          Rat Expression Set 230 1395313_s_at - -
          Rat Expression Set 230 1395313_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775628 - -
          Rat Genome U34 Set rc_AI639072_at - -
          Rat Genome U34 Set AA850219_at - -
          Rat Genome U34 Set rc_AA945932_g_at - -
          Rat Genome U34 Set rc_AA945932_at - -
          Rat Genome U34 Set rc_AI146189_s_at - -
          Rat Genome U34 Set rc_AI179982_s_at - -
          Rat Genome U34 Set rc_AI179982_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22000040 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368619 - -
          RatRef-12 Expression BeadChip ILMN_57519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0